Skip to Content
Merck
All Photos(1)

Key Documents

T2684

Sigma-Aldrich

TiterMax® Gold Adjuvant

liquid

Synonym(s):

TiterMax adjuvant

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

conjugate

unconjugated

Quality Level

form

liquid

availability

not available in Japan

storage temp.

2-8°C

General description

TiterMax Gold Adjuvant contains three essential ingredients: a new block copolymer, CRL-8300, squalene (a metabolizable oil) and a sorbitan monooleate.

Application

TiterMax® Gold Adjuvant has been used:
  • to emulsify soluble antigen-1 (Ag-1) to immunize animals to generate fully human monoclonal antibodies from XenoMouse strains
  • to inject GST-dGIPC (Drosophila homolog of GAIP interacting protein) protein into rats for generating antibodies against dGIPC protein
  • with Drosophila CtBP (dCtBP) protein in phosphate-buffered saline (PBS) to subcutaneously inject in a New Zealand female rabbit

Biochem/physiol Actions

Specific proteins can be emulsified with equal volume of TiterMax® Gold Adjuvant to immunize lab animals and extract antibodies from them.
TiterMax Gold is a new and improved water-in-oil adjuvant. By developing a new block copolymer and eliminating the silica from the formulation, TiterMax Gold appears to be less toxic and more effective than TiterMax Classic. The optimum volume of TiterMax Gold for an emulsion is twice that of TiterMax Classic.

Legal Information

TiterMax is a registered trademark of TiterMax USA, Inc

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

related product

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jihyun Kim et al.
Biochemical and biophysical research communications, 402(3), 565-570 (2010-10-30)
To identify genes that function in the adult neural system, we screened pools of P element-mediated mutants and tested locomotor activity of homozygous flies. Of 1014 P element-mutagenized lines, 638 were homozygous viable. These lines were tested for climbing ability
Elwira Pyz et al.
European journal of immunology, 38(4), 1157-1163 (2008-03-20)
Inhibitory receptors are required for the control of cellular activation and they play essential roles in regulating homeostasis and immunity. We previously identified a human inhibitory C-type lectin-like receptor, MICL (CLEC12A), a heavily glycosylated monomer predominantly expressed on myeloid cells.
Yuri Perepliotchikov et al.
Journal of virological methods, 260, 62-69 (2018-07-14)
Environmental surveillance (EnvS)2 is an important tool for monitoring the presence of poliovirus in endemic and poliovirus free regions. Unlike acute flaccid paralysis (AFP)3 surveillance, EnvS can monitor large populations using small numbers of samples and detect the introduction of
Paolo Struffi et al.
The Journal of biological chemistry, 280(49), 40757-40765 (2005-09-28)
Knirps and other short range transcriptional repressors play critical roles in patterning the early Drosophila embryo. These repressors are known to bind the C-terminal binding protein corepressor, but their mechanism of action is poorly understood. We purified functional recombinant Knirps
Anand Srivastava et al.
PloS one, 6(5), e20270-e20270 (2011-06-01)
Var2CSA, a key molecule linked with pregnancy-associated malaria (PAM), causes sequestration of Plasmodium falciparum infected erythrocytes (PEs) in the placenta by adhesion to chondroitin sulfate A (CSA). Var2CSA possesses a 300 kDa extracellular region composed of six Duffy-binding like (DBL)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service