Skip to Content
Merck
All Photos(1)

Key Documents

STS0025

Sigma-Aldrich

TERGITOL

new
greener alternative

Type 15-S-9, for molecular biology

Synonym(s):

Secondary alcohol ethoxylate

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12161900
NACRES:
NA.25

grade

for molecular biology

Quality Level

description

non-ionic
(application(s): life science and biopharma)

type

Type 15-S-9

form

liquid

mol wt

596 g/mol

greener alternative product characteristics

Design for Degradation
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

CMC

52 ppm

transition temp

cloud point 60 °C
pour point 9 °C

HLB

13.3

foreign activity

foreign activity (Dnase, Rnase, none detected)

greener alternative category

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Green Chemistry. This product is a biodegradable surfactant and is aligned with the 10th principle of Green Chemistry "Design for Degradation".

Tergitol 15-S-9 is a linear non-ionic surfactant that is a secondary ethoxylated alcohol. This clear liquid surfactant is compatible with anionic, cationic, and other nonionic surfactants.

Application

Tergitol has been used as a surfactant to study its influence on the formation of multi-phase micro-emulsions.
Tergitol™ 15-S-9 has been used:
  • To synthesize thermally stable mesoporous alumina
  • To synthesize porous TiO2
  • Along with SDS, ethanol and water for continuous phase preparations in electrospray emulsification
  • In the synthesis of Lath-like γ-alumina and boehmite nanofibers

Features and Benefits

  • Superior detergency
  • Rapid dissolution & good rinseability
  • Excellent formulation & handling properties
  • Narrow gel range
  • Readily biodegradable (OECD 301E)
  • Low odor

Other Notes

Polyglycol ether (nonionic) surfactants.

Legal Information

TERGITOL is a trademark of The Dow Chemical Company ("Dow") or an affiliated company of Dow

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pillared laponite clays-supported palladium catalysts for the complete oxidation of benzene.
Jinjun L, et al.
J. Mol. Catal. A: Chem., 225(2), 173-179 null
Interfacial tension and phase behavior of surfactant-brine?oil system.
Bera A, et al.
Colloids and Surfaces. A, Physicochemical and Engineering Aspects, 383(1-3), 114-119 (2011)
Preparation method and some tribological properties of porous titanium dioxide layers.
Piwonski I
Thin Solid Films, 515(7-8), 3499-3506 (2007)
Uniform discotic wax particles via electrospray emulsification.
Journal of Colloid and Interface Science, 334(1), 22-28 (2009)
Thermally stable mesoporous alumina synthesized with non-ionic surfactants in the presence of amines.
Gonzalez-Pena V, et al.
Microporous and Mesoporous Materials : The Official Journal of the International Zeolite Association, 44, 203-210 (2001)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service