Skip to Content
Merck
All Photos(1)

Key Documents

STR1

Sigma-Aldrich

Enhanced Avian First Strand Synthesis Kit

Components for cDNA synthesis with enhanced AMV reverse transcriptase

Synonym(s):

Reverse Transcriptase cDNA synthesis kit, eAMV-RT

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.55

usage

sufficient for 50 reactions
 kit sufficient for 50 reactions

feature

dNTPs included
hotstart: no

technique(s)

RT-PCR: suitable

color

colorless

input

purified RNA

shipped in

dry ice

storage temp.

−20°C

General description

Enhanced Avian First Strand Synthesis Kit utilizes a highly purified avian myeloblastosis virus reverse transcriptase (eAMV-RT) that offers superior performance in comparison to standard AMV-RT or standard Moloney murine leukemia virus reverse transcriptase (MMLV-RT). This exceptionally robust eAMV-RT has an enhanced ability to transcribe through difficult secondary structure at elevated temperatures (up to 65 °C) making it the ideal enzyme for producing high quality full-length cDNA from total RNA or poly(A)+ RNA.
Sigma′s Enhanced Avian RT First Strand Synthesis Kit also provides random nonamers (9-mers) and anchored oligo (dT)23 primers since gene-specific primers may not always be useful or possible. The anchored oligo has 23 thymidine residues and one G, C, or A residue (the anchor). The anchor ensures that the oligo (dT) primer binds at the very beginning of the message and that there is not a long region of the unusable sequence. With this kit, a dependable cDNA is generated that can be used for various downstream applications, including PCR.

Application

Enhanced Avian First Strand Synthesis Kit has been used for cDNA synthesis, which is intended for use in a real-time reverse transcription-polymerase chain reaction (RT-PCR).

Unit Definition

One unit incorporates one nanomole of TMP into TCA-precipitable material in 10 minutes using polyadenylic acid as template and oligo(dT)12-18 as primer.

Legal Information

Purchase of this product is accompanied by a limited license for use in the Polymerase Chain Reaction (PCR) process for research purposes only and in conjunction with a thermal cycler whose use in the automated performance of the PCR process is covered by an up-front license fee, either by payment to Applied Biosystems or as purchased, i.e., and authorized thermal cycler.

Kit Components Only

Product No.
Description

  • Enhanced Avian Reverse Transcriptase 1000 U

Kit Components Also Available Separately

Product No.
Description
SDS

  • Deoxynucleotide mix 50 μL

  • O4387Anchored oligo (dT)23 100 μLSDS

  • R7647Random nonamers 100 μLSDS

  • O4387Ribonuclease inhibitor 50 μLSDS

  • W1754PCR grade water 1.5 mLSDS

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Heat shock proteins expression during thermal risk exposure in the temperate xerothermic ant Formica cinerea
Slipinski P, et al.
Sociobiology, 62(3), 457-459 (2015)
Sigma-Aldrich Corporation's Life Science Quarterly. Hot Start RT-PCR results in improved performance of the enhanced Avian RT-PCR
Easlund, E., and Mueller, E.
Sigma data, 2-5 (2001)
JianFei Wang et al.
Journal of leukocyte biology, 82(6), 1554-1563 (2007-11-28)
Wound healing is a complex process involving the integrated actions of numerous cell types, soluble mediators, and ECM. Recently, a newly identified cell type, the fibrocyte, has been reported to contribute to wound healing and fibrotic conditions such as hypertrophic
Julio Carrion et al.
Journal of immunology (Baltimore, Md. : 1950), 189(6), 3178-3187 (2012-08-15)
The low-grade oral infection chronic periodontitis (CP) has been implicated in coronary artery disease risk, but the mechanisms are unclear. In this study, a pathophysiological role for blood dendritic cells (DCs) in systemic dissemination of oral mucosal pathogens to atherosclerotic
María José Martín et al.
The Journal of biological chemistry, 282(47), 34510-34524 (2007-09-21)
Heme oxygenase-1 (HO-1), an inducible enzyme that metabolizes the heme group, is highly expressed in human Kaposi sarcoma lesions. Its expression is up-regulated by the G protein-coupled receptor from the Kaposi sarcoma-associated herpes virus (vGPCR). Although recent evidence shows that

Articles

Challenges in gene expression analysis include mRNA stability, temporal transcription patterns, and mRNA-protein correlation, impacting accuracy.

Challenges in gene expression analysis include mRNA stability, temporal transcription patterns, and mRNA-protein correlation, impacting accuracy.

Challenges in gene expression analysis include mRNA stability, temporal transcription patterns, and mRNA-protein correlation, impacting accuracy.

Challenges in gene expression analysis include mRNA stability, temporal transcription patterns, and mRNA-protein correlation, impacting accuracy.

Protocols

The 3’/5’ integrity assay identifies RNA degradation, crucial for large sample sets or less detectable degradation.

The 3’/5’ integrity assay identifies RNA degradation, crucial for large sample sets or less detectable degradation.

Related Content

Polymerase chain reaction (PCR) is a technique for amplifying nucleic acid molecules and is commonly used in many applications, including RT-PCR, hot start PCR, end point PCR and more.

Polymerase chain reaction (PCR) is a technique for amplifying nucleic acid molecules and is commonly used in many applications, including RT-PCR, hot start PCR, end point PCR and more.

Polymerase chain reaction (PCR) is a technique for amplifying nucleic acid molecules and is commonly used in many applications, including RT-PCR, hot start PCR, end point PCR and more.

Polymerase chain reaction (PCR) is a technique for amplifying nucleic acid molecules and is commonly used in many applications, including RT-PCR, hot start PCR, end point PCR and more.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service