Skip to Content
Merck
All Photos(1)

Key Documents

SRP6303

Sigma-Aldrich

Apolipoprotein E from human plasma

≥95% (SDS-PAGE)

Synonym(s):

ApoE

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

biological source

human plasma

Assay

≥95% (SDS-PAGE)

form

frozen liquid

mol wt

34 kDa

packaging

pkg of 50 μg

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... ApoE(348)

General description

Apolipoprotein E (ApoE) is an important constituent of all plasma lipoproteins. The concentration of Apo E in normal plasma is 5 mg/100mL. Significant quantities of ApoE are produced in liver and brain and to some extent in almost every organ. It exists in three major isoforms; E2, E3, and E4, which differ from one another by a single amino-acid substitution. The gene encoding this protein is localized on human chromosome 19q13.32.

Biochem/physiol Actions

Apolipoprotein E (ApoE) serves as a ligand for low density receptors and participates in the transport and redistribution of cholesterol and other lipids. Other functions include immunoregulation, cell growth modulation and differentiation. Apo E is thought to be involved in tissue repair as increased amounts of the protein are found at sites of peripheral nerve injury and regeneration. A mutant form is associated with familial type III hyperlipoproteinemia. In addition to facilitating solubilization of lipids, these proteins help to maintain the structural integrity of lipoproteins, serve as ligands for lipoprotein receptors, and regulate the activity of enzymes involved in lipid metabolism. ApoE′s interaction with specific ApoE receptor enables uptake of chylomicron remnants by liver cells, which is an essential step during normal lipid metabolism. It also binds with the low-density lipoprotein (LDL) receptor (apo B/E).

Physical form

Frozen in 50 mM NH4HCO3, pH 8.0.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Genome-wide association study identifies a single major locus contributing to survival into old age; the APOE locus revisited.
Deelen J
Aging Cell, 10(4), 686-698 (2011)
Molecular mechanism of apolipoprotein E binding to lipoprotein particles.
Nguyen D
Biochemistry, 48(13), 3025-3032 (2009)
Pathogenesis of type III hyperlipoproteinemia (dysbetalipoproteinemia). Questions, quandaries, and paradoxes.
Mahley RW
Journal of Lipid Research, 40(11), 1933-1949 (1999)
Apolipoprotein E-low density lipoprotein receptor binding: study of protein-protein interaction in rationally selected docked complexes.
Prevost M and Raussens V
Proteins: Structure, Function, and Genetics, 55(4), 874-884 (2004)
Apolipoprotein E isoforms and lipoprotein metabolism.
Phillips MC
IUBMB Life, 66(9), 616-623 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service