Skip to Content
Merck
All Photos(1)

Key Documents

SRP3201

Sigma-Aldrich

GM-CSF from mouse

Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture

Synonym(s):

CSF-2, MGI-1GM, pluripoietin-?

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

biological source

mouse

recombinant

expressed in E. coli

Assay

≥98% (HPLC)
≥98% (SDS-PAGE)

form

lyophilized

potency

≤0.05 ng/mL

mol wt

14.2 kDa

packaging

pkg of 20 μg

technique(s)

cell culture | mammalian: suitable

impurities

<0.1 EU/μg endotoxin, tested

color

white to off-white

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

mouse ... CSF2(12981)

General description

GM-CSF (granulocyte-macrophage colony-stimulating factor, also referred to as CSF2) is produced in by endothelial cells, monocytes, fibroblasts and T-lymphocytes. Recombinant murine GM-CSF is a 14.2kDa globular protein consisting of 124 amino acids residues. The human and murine molecules are species-specific and exhibit no cross-species reactivity.

Application

GM-CSF (granulocyte-macrophage colony-stimulating factor) mouse has been used to check effect of GM-CSF on retinal ganglion cell neurite outgrowth in the retinal explants.

Biochem/physiol Actions

GM-CSF (granulocyte-macrophage colony-stimulating factor, also referred to as CSF2) is a hematopoietic growth factor that stimulates the development of neutrophils and macrophages and promotes the proliferation and development of early erythroid megakaryocytic and eosinophilic progenitor cells. It enhances neutrophil migration. GM-CSF plays a crucial role in inflammatory and autoimmune diseases. In neutrophils, it promotes the expression of TLR2 (Toll-like receptor) and TLR2-associated interleukin 8 responses, thereby eliminating microbial attack.

Sequence

MAPTRSPITV TRPWKHVEAI KEALNLLDDM PVTLNEEVEV VSNEFSFKKL TCVQTRLKIF EQGLRGNFTK LKGALNMTAS YYQTYCPPTP ETDCETQVTT YADFIDSLKT FLTDIPFECK KPVQK

Physical form

Lyophilized from 10 mM acetic acid

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in water to a concentration of 0.1-1.0 mg/ml. Do not vortex. This solution can be stored at 2-8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a stabilizer (example 5% Trehalose) and store in working aliquots at -20°C to -80°C.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

GM-CSF in inflammation and autoimmunity.
Hamilton JA
Trends in Immunology, 23, 403-408 (2002)
Preparation of embryonic retinal explants to study CNS neurite growth.
Hanea ST
Experimental Eye Research, 146, 304-312 (2016)
Xuexia Ji et al.
Nan fang yi ke da xue xue bao = Journal of Southern Medical University, 40(4), 525-530 (2020-09-09)
To investigate the molecular mechanism underlying the inhibitory effect of propofol on pyroptosis of macrophages. Macrophages derived from bone marrow were extracted and divided into three groups: control group, LPS+ATP group and propofol+LPS+ATP group. The control group was not given
Role of toll-like receptor 2 (TLR2) in neutrophil activation: GM-CSF enhances TLR2 expression and TLR2-mediated interleukin 8 responses in neutrophils.
Kurt-Jones EA
Blood, 100, 1860-1868 (2002)
Na Li et al.
Bioengineered, 12(2), 10971-10981 (2021-11-06)
Macrophage infiltration is a hallmark pathological change observed in early stage myocardial ischemia/reperfusion (MI/R) injury and one of the main causes of myocardial damage. Here, we investigated the effects of p-Coumaric acid (p-CA) on macrophage polarization following MI/R injury and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service