Skip to Content
Merck
All Photos(1)

Key Documents

SRP3196

Sigma-Aldrich

EGF from mouse

recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture

Synonym(s):

Epidermal Growth Factor, URG, Urogastrone

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

biological source

mouse

recombinant

expressed in E. coli

Assay

≥98% (HPLC)
≥98% (SDS-PAGE)

form

lyophilized

potency

0.1 ng/mL

mol wt

6.0 kDa

packaging

pkg of 500 μg

technique(s)

cell culture | mammalian: suitable

impurities

<0.1 EU/μg endotoxin, tested

color

off-white to yellow

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

mouse ... EGF(13645)

General description

EGF (epidermal growth factor) is a potent growth factor that stimulates the proliferation of various epidermal and epithelial cells. Recombinant murine EGF is a 6kDa globular protein containing 53 amino acid residues, including 3 intramolecular disulfide-bonds.

Application

EGF from mouse has also been used in the DMEM/F12 media for growing mouse kidney cells.

Biochem/physiol Actions

EGF (epidermal growth factor) has been shown to inhibit gastric secretion, and to be involved in wound healing. EGF signals through a receptor known as c-erbB, which is a class I tyrosine kinase receptor. This receptor also binds with TGF (transforming growth factor)-α and VGF (vaccinia virus growth factor). EGF plays an important role in embryogenesis and cardiogenesis.

Sequence

NSYPGCPSSY DGYCLNGGVC MHIESLDSYT CNCVIGYSGD RCQTRDLRWW ELR

Physical form

Lyophilized from 10 mM Sodium Phosphate, pH 8.2.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in water to a concentration of 0.5-1.0 mg/ml. Do not vortex. This solution can be stored at 2-8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein (example 0.1% BSA) and store in working aliquots at -20°C to -80°C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

EGF is required for cardiac differentiation of P19CL6 cells through interaction with GATA-4 in a time- and dose-dependent manner.
Ma CX
Cellular and Molecular Life Sciences, 72, 2005-2022 (2015)
Requirement of Leukemia Inhibitory Factor or Epidermal Growth Factor for Pre-Implantation Embryogenesis via JAK/STAT3 Signaling Pathways.
Cheng EH
PLoS ONE, 11, e0153086-e0153086 (2016)
Fatty Acid Oxidation is Impaired in An Orthologous Mouse Model of Autosomal Dominant Polycystic Kidney Disease.
Menezes LF
EBioMedicine, 5, 183-192 (2016)
Epidermal growth factor therapy and wound healing--past, present and future perspectives.
Hardwicke J
The Surgeon : Journal of the Royal Colleges of Surgeons of Edinburgh and Ireland, 6, 172-177 (2008)
Transforming growth factor alpha and epidermal growth factor in protection and healing of gastric mucosal injury.
Konturek SJ
Scandinavian Journal of Gastroenterology, 27, 649-655 (1992)

Articles

Organoid culture products to generate tissue and stem cell derived 3D brain, intestinal, gut, lung and cancer tumor organoid models.

Organoid culture products to generate tissue and stem cell derived 3D brain, intestinal, gut, lung and cancer tumor organoid models.

Organoid culture products to generate tissue and stem cell derived 3D brain, intestinal, gut, lung and cancer tumor organoid models.

Organoid culture products to generate tissue and stem cell derived 3D brain, intestinal, gut, lung and cancer tumor organoid models.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service