SRP3147
Relaxin-2 Peptide
≥98% (HPLC), human recombinant, expressed in E. coli, suitable for cell culture
Synonym(s):
H2 relaxin, RLN2
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352202
NACRES:
NA.32
Recommended Products
Product Name
Relaxin-2 human, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
biological source
human
recombinant
expressed in E. coli
Assay
≥98% (HPLC)
≥98% (SDS-PAGE)
form
lyophilized
mol wt
6.0 kDa
packaging
pkg of 25 μg
technique(s)
cell culture | mammalian: suitable
impurities
<0.1 EU/μg endotoxin, tested
color
white
UniProt accession no.
shipped in
wet ice
storage temp.
−20°C
Gene Information
human ... RLN2(6019)
Related Categories
General description
Relaxin-2 is a peptide hormone structurally related to insulin. The gene is mapped to human chromosome 9p24. It is expressed in the placenta, decidua, prostate, corpus luteum, endometrium, mammary glands, heart, and brain. It is also expressed in the ovary during pregnancy. Recombinant relaxin-2 is a nonglycosylated 6kDa disulfide linked heterodimeric protein consisting of a 24 amino acid A-chain and a 29 amino acid B-chain.
Application
It has also been used to study its effect on human endometrial carcinoma cell lines.
Relaxin-2 human has been used to study the effect of testosterone on the expression of relaxin receptor in the knee joint.
Biochem/physiol Actions
Of the three known relaxin genes, Relaxin-2 (RLN2) is the only relaxin known to circulate in the blood. Relaxin-2 binds specifically to the LGR7 (leucine-rich repeat-containing G-protein coupled receptor 7) and LGR8 receptors, previously identified as an "orphan" G protein coupled receptors. Signaling by Relaxin-2 through its target receptors enhances the growth of pubic ligaments and ripening of the cervix during birth. Relaxin-2 also participates in cancer progression. In prostate cancer, it is responsible for tumor growth and vascularisation. Relaxin-2 is also linked with anti-fibrotic and cardioprotective effects.
Sequence
A-Chain: QLYSALANKC CHVGCTKRSL ARFC B-Chain: DSWMEEVIKL CGRELVRAQI AICGMSTWS
Physical form
Lyophilized with no additives.
Reconstitution
Centrifuge the vial prior to opening. Reconstitute in water to a concentration of 0.1-1.0 mg/ml. Do not vortex. This solution can be stored at 2-8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein (example 0.1% BSA) and store in working aliquots at -20°C to -80°C.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Dual blockade of PKA and NF-?B inhibits H2 relaxin-mediated castrate-resistant growth of prostate cancer sublines and induces apoptosis.
Vinall RL, et al.
Hormones & Cancer, 2, 224-238 (2011)
Human relaxin-2: historical perspectives and role in cancer biology.
Nair VB, et al.
Amino Acids, 43, 1131-1140 (2012)
DNA sequence and analysis of human chromosome 9.
Humphray SJ, et al.
Nature, 429, 369-374 (2004)
Relaxin 2/RXFP1 Signaling Induces Cell Invasion via the beta-Catenin Pathway in Endometrial Cancer
Fue M, et al.
International Journal of Molecular Sciences, 19(8), 2438-2438 (2018)
Relaxin as a cardiovascular hormone: physiology, pathophysiology and therapeutic promises.
Nistri S, et al.
Cardiovascular & Hematological Agents in Medicinal Chemistry, 5, 101-108 (2007)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service