Skip to Content
Merck
All Photos(1)

Key Documents

SRP3084

Sigma-Aldrich

IL-1Ra human

recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture

Synonym(s):

ICIL-1RA, IL-1RN, IRAP

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

recombinant

expressed in E. coli

Assay

≥98% (HPLC)
≥98% (SDS-PAGE)

form

lyophilized

potency

20-40 ng/mL

mol wt

17.2 kDa

packaging

pkg of 100 μg

technique(s)

cell culture | mammalian: suitable

impurities

<0.1 EU/μg endotoxin, tested

color

white to off-white

suitability

suitable for molecular biology

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... IL1R1(3554)

General description

Research Area: IMMUNO AND CKS
The interleukin-1 receptor antagonist (IL-1Ra) is part of the IL-1 family and binds to IL-1 receptors without triggering any intracellular response. IL-1Ra, produced by hepatic cells, exhibits characteristics of an acute phase protein. Endogenous IL-1Ra is generated in various experimental animal models of disease, as well as in human autoimmune and chronic inflammatory conditions.
The gene IL-1RA (interleukin-1 receptor antagonist) is mapped to human chromosome 2q. It belongs to the IL-1 (interleukin 1) family of proteins. The encoded protein has a TIR (Toll/IL-1 receptor) domain. IL-1RA is produced by corneal epithelial cells, monocytes, neutrophils, macrophages and fibroblasts. It is a naturally occurring inflammatory inhibitor protein. Recombinant human IL-1RA is a 17.2kDa protein consisting of 153 amino acid residues.

Application

IL-1Ra human has been used as an IL1 antagonist to block the signaling from the IL1 receptor and verify that IL1 signaling is responsible for invasiveness in CD133-expressing cells. It has also been used as a control in sandwich enzyme-linked immunosorbent assay (ELISA) to measure protein levels.

Biochem/physiol Actions

Interleukin-1 receptor antagonist (IL-1RA) inhibits the activity of IL-1α (interleukin 1 α) and IL-1β (interleukin 1 β) by competitively blocking binding to their type I and type II receptors. Therapeutically, IL-1RA may help in the treatment of sepsis, rheumatoid arthritis, chronic myelogenous leukemia, asthma, psoriasis, and inflammatory bowel disease. Low production of IL-1RA is observed in HIV-1 (human immunodeficiency virus 1)-infected dendritic cells.
The interleukin-1 receptor antagonist (IL-1Ra) functions as a specific and selective competitive inhibitor of IL-1R1, binding to the receptor with high affinity.

Sequence

MRPSGRKSSK MQAFRIWDVN QKTFYLRNNQ LVAGYLQGPN VNLEEKIDVV PIEPHALFLG IHGGKMCLSC VKSGDETRLQ LEAVNITDLS ENRKQDKRFA FIRSDSGPTT SFESAACPGW FLCTAMEADQ PVSLTNMPDE GVMVTKFYFQ EDE

Physical form

Lyophilized from 5mM Tris, pH 8.0 + 25 mM NaCl.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in water to a concentration of 0.1-1.0mg/ml. Do not vortex. This solution can be stored at 2-8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein (example 0.1% BSA) and store in working aliquots at -20°C to -80°C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Therapies negating neuroinflammation after brain trauma
Hellewell S, et al.
Brain Research, 36-56 (2016)
Identification of critical regions within the TIR domain of IL-1 receptor type I.
Radons J
The International Journal of Biochemistry & Cell Biology, 68, 15-20 (2015)
The effects of interleukin-10 on interleukin-1 receptor antagonist and interleukin-1 beta production in human monocytes and neutrophils.
Jenkins JK
Lymphokine and Cytokine Research, 13, 47-54 (1994)
A synthetic mammalian therapeutic gene circuit for sensing and suppressing inflammation
Smole A, et al.
Molecular Therapy, 102-119 (2017)
Association between interleukin-1 receptor antagonist (IL-1ra) gene polymorphism and early and late-onset psoriasis.
Tarlow JK
British Journal of Dermatology, 136, 147-148 (1997)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service