SRP3071
IL-10 human
recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Synonym(s):
B-TCGF, CSIF, TGIF
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352202
NACRES:
NA.32
Recommended Products
biological source
human
recombinant
expressed in E. coli
Assay
≥98% (HPLC)
≥98% (SDS-PAGE)
form
lyophilized
potency
≤2.0 ng/mL ED50
mol wt
18.6 kDa
packaging
pkg of 10 μg
technique(s)
cell culture | mammalian: suitable
impurities
<0.1 EU/μg endotoxin, tested
color
white to off-white
UniProt accession no.
shipped in
wet ice
storage temp.
−20°C
Gene Information
human ... IL10(3586)
General description
IL-10 (interleukin 10) is an immunosuppressive cytokine produced by a variety of mammalian cell types including macrophages, monocytes, T cells, B cells and keratinocytes. The gene is mapped to human chromosome 1q31-1q32. Recombinant human IL-10 is an 18.6kDa protein of 161 amino acid residues. Human IL-10 active on murine cells, but murine IL-10 is inactive on human cells.
Application
IL-10 human has been used to study the effect of IL-10 on endothelial nitric oxide synthase and nitric oxide in African American human umbilical vein endothelial cells. It has also been used to study the effect of IL-10 on leukotriene (LT) A4 hydrolase and LTC4 synthase protein expression in and secretion from, an inflamed porcine endometrium.
Biochem/physiol Actions
IL-10 (interleukin 10) is an anti-inflammatory cytokine. IL-10 inhibits the expression of proinflammatory cytokines such as IL-1 and TNF (tumor necrosis factor)-α. Like IL-4, IL-10 enhances humoral immune responses and attenuates cell-mediated immune reactions. Polymorphisms in the IL-10 gene are associated with higher risk of preeclampsia, ankylosing spondylitis, nasopharyngeal carcinoma and basal cell carcinoma. Polymorphism in IL-10 at -1082A/G is associated with diabetic nephropathy.
Sequence
MSPGQGTQSE NSCTHFPGNL PNMLRDLRDA FSRVKTFFQM KDQLDNLLLK ESLLEDFKGY LGCQALSEMI QFYLEEVMPQ AENQDPDIKA HVNSLGENLK TLRLRLRRCH RFLPCENKSK AVEQVKNAFN KLQEKGIYKA MSEFDIFINY IEAYMTMKIR N
Physical form
Lyophilized from 5 mM Sodium Phosphate, pH 7.2.
Reconstitution
Centrifuge the vial prior to opening. Reconstitute in 5 mM Sodium Phosphate, pH 7.2 to a concentration of 0.1-1.0 mg/ml. Do not vortex. This solution can be stored at 2-8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein (example 0.1% BSA) and store in working aliquots at -20°C to -80°C.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Interleukin-10 polymorphisms and nasopharyngeal carcinoma risk: a meta-analysis.
Yu YF, et al.
Genetics and molecular research : GMR, 14, 18945-18957 (2015)
Effect of Interleukin-10 and Laminar Shear Stress on Endothelial Nitric Oxide Synthase and Nitric Oxide in African American Human Umbilical Vein Endothelial Cells.
Babbitt DM, et al.
Ethnicity & Disease, 25, 413-418 (2015)
TNF-a and IL10 polymorphisms interaction increases the risk of ankylosing spondylitis in Chinese Han population.
Wang NG, et al.
International Journal of Clinical and Experimental Pathology, 8, 15204-15209 (2015)
Messenger RNA expression of the immunosuppressive cytokine IL-10 in human gliomas.
Huettner C, et al.
The American Journal of Pathology, 146, 317-322 (1995)
Investigation on the association between inerleukin-10 -592C/A, 819C/T and -1082A/G gene polymorphisms and development of diabetic nephrophathy.
Yin Q, et al.
International Journal of Clinical and Experimental Pathology, 8, 15216-15221 (2015)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service