Skip to Content
Merck
All Photos(1)

Key Documents

SRP0417

Sigma-Aldrich

BirA

recombinant, expressed in E. coli, ≥65% (SDS-PAGE)

Synonym(s):

BirA Biotin Ligase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352204
NACRES:
NA.32

recombinant

expressed in E. coli

Assay

≥65% (SDS-PAGE)

form

aqueous solution

mol wt

37 kDa

packaging

pkg of 100 μg

NCBI accession no.

shipped in

dry ice

storage temp.

−70°C

General description

Biotin ligase BirA (GenBank Accession No. AP012306 (3602298-3603260)) amino acids 2 – 321 (end) with His-FLAG-tag, MW = 37.1 kDa, expressed in an Escherichia coli cell expression system.

Application

Useful for the study of enzyme kinetics, screening inhibitors, and selectivity profiling.

Biochem/physiol Actions

Biotin ligase BirA controls biotin synthesis in Escherichia coli. It sends biotin to metabolism. As a homodimer, it negatively controls the biotin synthetic operon. BirA also catalyzes post-translational biotinylation, by associating with biotin acceptor protein of acetyl-coenzyme A carboxylase. It can biotinylate human histones.

Physical form

Formulated in 50 mM Tris-HCl, pH 8.0, 50 mM NaCl, 150 mM imidazole, 3 mM DTT, and 5% Glycerol.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Prokaryotic BirA ligase biotinylates K4, K9, K18 and K23 in histone H3.
Kobza K, et al.
Bmb Reports, 41, 310-315 (2008)
Vandana Chakravartty et al.
Journal of bacteriology, 194(5), 1113-1126 (2012-01-03)
Transcription of the Escherichia coli biotin (bio) operon is directly regulated by the biotin protein ligase BirA, the enzyme that covalently attaches biotin to its cognate acceptor proteins. Binding of BirA to the bio operator requires dimerization of the protein
Functional versatility of a single protein surface in two protein:protein interactions.
Adikaram PR and Beckett D
Journal of Molecular Biology, 419, 223-233 (2012)
Bin Yang et al.
Nature communications, 6, 6592-6592 (2015-03-21)
Enterohemorrhagic Escherichia coli (EHEC) is an important foodborne pathogen that infects humans by colonizing the large intestine. Here we identify a virulence-regulating pathway in which the biotin protein ligase BirA signals to the global regulator Fur, which in turn activates
Yifeng Li et al.
Protein expression and purification, 82(1), 162-167 (2012-01-10)
The extremely tight binding between biotin and avidin or streptavidin makes labeling proteins with biotin a useful tool for many applications. BirA is the Escherichia coli biotin ligase that site-specifically biotinylates a lysine side chain within a 15-amino acid acceptor

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service