SRP0158
Histone H3 (2-58) human
recombinant, expressed in E. coli, ≥70% (SDS-PAGE)
Synonym(s):
HIST1H3E, Histone H3.1
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
biological source
human
recombinant
expressed in E. coli
Assay
≥70% (SDS-PAGE)
form
aqueous solution
mol wt
32 kDa
packaging
pkg of 500 μg
storage condition
avoid repeated freeze/thaw cycles
concentration
>0.02 mg/mL
NCBI accession no.
UniProt accession no.
shipped in
dry ice
storage temp.
−70°C
Gene Information
human ... HIST1H3E(8353)
General description
The mammalian chromatin is present in nucleosomes, made of histone octamers (H2A,H2B,H3,H4)2. Mammals contain three main classes of histone H3 variants: the replicative histones (H3.1 and H3.2), the replacement histone (H3.3) and the centromeric histone (Cenp-A).
Human Histone 3 (GenBank Accession No. NM_003532), (amino acid 2-58) with N-terminal GST tag, MW = 32kDa, expressed in an Escherichia coli expression system.
Human Histone 3 (GenBank Accession No. NM_003532), (amino acid 2-58) with N-terminal GST tag, MW = 32kDa, expressed in an Escherichia coli expression system.
Application
Suitable substrate for histone methyltransferases
Biochem/physiol Actions
Histone proteins are basic building blocks of chromatin. H3.1 (also referred to as HIST1H3E) is mainly expressed in the S phase and is termed as replication-dependent histone. Selective methylation of H3.1 controls replication in heterochromatin area.
Physical form
Formulated in 25 mM Tris-HCl, pH 8.0, 100 mM NaCl, 0.05% Tween-20, 20% glycerol and 3 mM DTT.
Preparation Note
Thaw on ice. Upon first thaw, briefly spin tube containing enzyme to recover full content of the tube. Aliquot enzyme into single use aliquots. Store remaining undiluted enzyme in aliquots at -70°C. Note: Enzyme is very sensitive to freeze/thaw cycles.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Selective methylation of histone H3 variant H3.1 regulates heterochromatin replication.
Science, 343, 1249-1249 (2014)
Genome-wide analysis of histone H3 lysine 4 trimethylation in peripheral blood mononuclear cells of minimal change nephrotic syndrome patients.
American Journal of Nephrology, 30, 505-505 (2009)
Topography of the histone octamer surface: repeating structural motifs utilized in the docking of nucleosomal DNA.
Proceedings of the National Academy of Sciences of the USA, 90, 10489-10489 (1993)
Nature structural & molecular biology, 17(11), 1343-1351 (2010-10-19)
The mechanism by which newly synthesized histones are imported into the nucleus and deposited onto replicating chromatin alongside segregating nucleosomal counterparts is poorly understood, yet this program is expected to bear on the putative epigenetic nature of histone post-translational modifications.
Signaling to chromatin through histone modifications.
Cell, 103(2), 263-271 (2000-11-01)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service