SRE0117
Salt Active Nuclease (SAN)
High Purity, recombinant, expressed in Pichia pastoris, Triton-free
Synonym(s):
endonuclease, salt active endonuclease, salt tolerant nuclease
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352204
NACRES:
NA.54
recombinant
expressed in Pichia pastoris
Quality Level
composition
Triton-free
packaging
pkg of 25 kU
shipped in
dry ice
storage temp.
−20°C
Looking for similar products? Visit Product Comparison Guide
General description
This Salt Active Nuclease (SAN) is the ultimate solution for efficient removal of nucleic acids in manufacturing and bioprocessing workflows. This nonspecific, recombinant endonuclease has optimum activity at high salt concentrations, which can improve efficiency and yield in various workflows.
Application
- Useful for purification of tagged recombinant proteins
- Manufacturing of nucleic acid-free molecular enzymes and reagents
- Research of vaccines, viruses and antibodies
Features and Benefits
- Active at low temperatures
- Binds strongly to cation exchange media because of high isoelectric point, even at elevated pH and salt concentrations where few other proteins show binding
- Typically does not bind to anion exchange or IMAC media
- Detergent free No protease detected
Unit Definition
One unit is defined as the amount of enzyme that causes a ΔA260 = 1.0 in 30 minutes at 37 °C in 25 mM Tris-HCI pH 8.5 (@25 °C), 5 mM MgCl2, 500 mM NaCl, and 50 µg/mL calf thymus DNA.
Reconstitution
In solution
Other Notes
- Temperature: 5 – 40°C, optimal: ~35°C
- Salt concentration (NaCl / KCl): 150 - 900 mM, optimal: 400 - 650 mM
- Mg2+: >1 mM is required for activity, optimal 5 - 50 mM
- pH: 7.3 – 10.0, optimal 8.2 - 9.2
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service