Skip to Content
Merck
All Photos(1)

Key Documents

SML4075

Sigma-Aldrich

Ginsenoside Rg1

≥98% (HPLC)

Synonym(s):

(3β,6α,12β)-3,12-Dihydroxydammar-24-ene-6,20-diyl bis-β-D-glucopyranoside, Ginsenoside A2, Ginsenoside g1, Panaxoside A, Panaxoside Rg1, Panaxsaponin Rg1, Rg1 ginsenoside, Sanchinoside C1, Sanchinoside Rg1

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C42H72O14
CAS Number:
Molecular Weight:
801.01
MDL number:
UNSPSC Code:
12352200

biological source

Panax notoginseng (Burk.) F.H.Chen

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: soluble mg/mL, clear

storage temp.

-10 to -25°C

SMILES string

O1[C@H]([C@@H]([C@H]([C@@H]([C@H]1CO)O)O)O)O[C@@H]2[C@@H]3[C@@]([C@@H]4[C@]([C@]5([C@H]([C@@H](C4)O)[C@H](CC5)[C@@](O[C@@H]6O[C@@H]([C@H]([C@@H]([C@H]6O)O)O)CO)(CCC=C(C)C)C)C)(C2)C)(CC[C@@H](C3(C)C)O)C

InChI key

YURJSTAIMNSZAE-HHNZYBFYSA-N

Biochem/physiol Actions

Major component of Panax ginseng that exhibits a variety of pharmacological actions including regulation of organ function regulation of adult stem cells, inhibition of autophagy and endoplasmic reticulum stress, antioxidant, anti-inflammatory, and metabolic modulation.
Ginsenoside Rg1, a key component of Panax ginseng, is a tetracyclic triterpenoid derivative that displays various pharmacological properties such as regulating organ function, regulating adult stem cells, inhibiting autophagy and endoplasmic reticulum stress to protect cardiac function, antioxidant, anti-inflammatory, and metabolic modulation. Ginsenoside Rg1 has also been found to be an effective regulator of mesenchymal stem cells and to have a positive effect on hepatic steatosis.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qian Xie et al.
Frontiers in pharmacology, 14, 1179705-1179705 (2023-09-25)
Background: According to existing laboratory data, ginsenoside Rg1 may help cure diabetes and its complications by reducing oxidative stress (OS) and managing inflammation. However, this conclusion lacks reliability and is unclear. As a result, the purpose of this systematic review
Huimin Zhou et al.
Journal of ethnopharmacology, 324, 117794-117794 (2024-01-21)
Ginseng (Panax ginseng C. A. Meyer) is a precious traditional Chinese medicine with multiple pharmacological effects. Ginsenoside Rg1 is a main active ingredient extracted from ginseng, which is known for its age-delaying and antioxidant effects. Increasing evidence indicates that Rg1
Jun Guo et al.
Journal of leukocyte biology, 112(5), 1065-1077 (2022-07-02)
Acute kidney injury (AKI) represents a prevailing complication of sepsis, and its onset involves ferroptosis. Ginsenoside Rg1 exerts a positive effect on kidney diseases. This study explored the action of ginsenoside Rg1 in sepsis-induced AKI (SI-AKI) by regulating ferroptosis in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service