Skip to Content
Merck
All Photos(1)

Key Documents

SML3903

Sigma-Aldrich

Insulin glargine

≥95% (HPLC)

Synonym(s):

HOE 901, [GlyA21,ArgB31,ArgB32]insulin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C267H404N72O78S6
CAS Number:
Molecular Weight:
6062.89
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥95% (HPLC)

form

(Powder or Lyophilized powder or film)

storage condition

desiccated

color

white to off-white

storage temp.

-10 to -25°C

Biochem/physiol Actions

Insulin glargine is an approved long-acting insulin analog used for management of Diabetes Type 1 and Type 2. Insulin glargine is a human insulin analog where A-chain asparagine 21 was replaced by glycine and B-chain carboxy terminal was extended by two arginine residues.

Caution

Hygroscopic

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Growth promoting and metabolic activity of the human insulin analogue [GlyA21,ArgB31,ArgB32]insulin (HOE 901) in muscle cells
European Journal of Pharmacology, 320(2-3), 259-265 (1997)
Insulin glargine: a systematic review of a long-acting insulin analogue
Clinical Therapeutics, 25(6), 1541-1577 (2003)
Twenty Years of Insulin Gla-100: A Systematic Evaluation of Its Efficacy and Safety in Type 2 Diabetes Mellitus
Diabetes therapy : research, treatment and education of diabetes and related disorders, 13(8), 1409-1481 (2022)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service