Skip to Content
Merck
All Photos(1)

Key Documents

SML3080

Sigma-Aldrich

C14PC

≥98% (HPLC)

Synonym(s):

2-[[Hydroxy(tetradecyloxy)phosphinyl]oxy]-N,N,N-trimethyl- ethanaminium inner salt, CDI 166493, D 19390, FOS 14, Fos-choline 14, Tetradecylphosphocholine, n-Tetradecyl-phosphocholine, n-Tetradecylphosphocholine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H42NO4P
CAS Number:
Molecular Weight:
379.51
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

[O-]P(OCCCCCCCCCCCCCC)(OCC[N+](C)(C)C)=O

InChI

1S/C19H42NO4P/c1-5-6-7-8-9-10-11-12-13-14-15-16-18-23-25(21,22)24-19-17-20(2,3)4/h5-19H2,1-4H3

InChI key

BETUMLXGYDBOLV-UHFFFAOYSA-N

Biochem/physiol Actions

n-Tetradecylphosphocholine (C14PC), a more stable analog of myristoyl lysophosphatidylcholine, is an inhibitor of MARSA virulence. C14PC is a broad-spectrum inhibitor of Panton–Valentine leucocidin (PVL), LukED and α−toxin. C14PC protects primary human immune cells in vitro against cytolysis by PVL and α-toxin. C14PC can be used as a detergent for membrane solubilization.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jie Liu et al.
The Journal of biological chemistry, 295(18), 5944-5959 (2020-03-18)
The rapid emergence and dissemination of methicillin-resistant Staphylococcus aureus (MRSA) strains poses a major threat to public health. MRSA possesses an arsenal of secreted host-damaging virulence factors that mediate pathogenicity and blunt immune defenses. Panton-Valentine leukocidin (PVL) and α-toxin are
Danyang Guo et al.
Metabolism: clinical and experimental, 107, 154232-154232 (2020-04-18)
Parasympathetic nerve (PN) signaling plays a crucial role in the maintenance of pancreatic β-cell volume density (Vβ). PN may be pathologically affected in diabetic polyneuropathy (DPN). However, the association between the reduction of PNs in islets and Vβ and the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service