SML3034
Tolterodine L-tartrate
≥98% (HPLC)
Synonym(s):
2-[(1R)-3-[Bis(1-methylethyl)amino]-1-phenylpropyl]-4-methyl-phenol tartrate, Detrusitol, Kabi-2234, PF-346634, PNU-200583, PNU-200583E, UK-233743, s (R)-2-[3[bis(1-methylethyl)-amino]1-phenylpropyl]-4-methylphenol [R-(R*,R*)]2,3dihydroxybutanedioate (1:1) (salt)
About This Item
Quality Level
Assay
≥98% (HPLC)
form
powder
optical activity
[α]/D 26.0 to 32.0°, c = 1 in methanol
color
white to beige
solubility
DMSO: 2 mg/mL, clear
storage temp.
room temp
SMILES string
O[C@H]([C@@H](O)C(O)=O)C(O)=O.CC(C)N(CC[C@H](c1ccccc1)c2cc(C)ccc2O)C(C)C
InChI
1S/C22H31NO.C4H6O6/c1-16(2)23(17(3)4)14-13-20(19-9-7-6-8-10-19)21-15-18(5)11-12-22(21)24;5-1(3(7)8)2(6)4(9)10/h6-12,15-17,20,24H,13-14H2,1-5H3;1-2,5-6H,(H,7,8)(H,9,10)/t20-;1-,2-/m11/s1
InChI key
TWHNMSJGYKMTRB-KXYUELECSA-N
Looking for similar products? Visit Product Comparison Guide
Biochem/physiol Actions
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 2 - Repr. 2
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service