Skip to Content
Merck
All Photos(1)

Key Documents

SML3012

Sigma-Aldrich

Dovitinib

≥98% (HPLC)

Synonym(s):

4-Amino-5-fluoro-3-[6-(4-methyl-1-piperazinyl)-1H-benzimidazol-2-yl]-2(1H)-quinolinone, CHIR 258, CHIR-258, CHIR258, TKI 258, TKI-258, TKI258

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H21FN6O
CAS Number:
Molecular Weight:
392.43
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C1NC2=C(C(N)=C1C3=NC4=CC=C(N5CCN(CC5)C)C=C4N3)C(F)=CC=C2

InChI

1S/C21H21FN6O/c1-27-7-9-28(10-8-27)12-5-6-14-16(11-12)25-20(24-14)18-19(23)17-13(22)3-2-4-15(17)26-21(18)29/h2-6,11H,7-10H2,1H3,(H,24,25)(H3,23,26,29)

InChI key

PIQCTGMSNWUMAF-UHFFFAOYSA-N

Biochem/physiol Actions

Dovitinib (CHIR-258) is an orally active and potent receptor tyrosine kinase (RTK) inhibitor against class III (FLT3/KIT/PDGFR1/CSFR/PDGFR2 IC50 = 1/2/27/36/200 nM), class IV (FGFR1/3 IC50 = 8/9 nM), and class V (VEGFR1/2/3 IC50 = 10/13/8 nM) RTKs, but not INSR, EGFR1, ErbB2 or Raf (IC50 = 2.1, 2.2, >20, >25 μM, respectively). Dovitinib inhibits target RTKs phosphorylation in cultured cells (pVEGFR1 and pPDGFRβ IC90 <50 nM; KM12L4a) and exhibits in vivo anti-tumor efficacy in mice bearing KM12L4a or HCT116 human cancer xenografts (10-120 mg/kg/day p.o.).

Storage Class Code

11 - Combustible Solids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiaohua Xin et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 12(16), 4908-4915 (2006-08-18)
The ectopically expressed and deregulated fibroblast growth factor receptor 3 (FGFR3) results from a t(4;14) chromosomal translocation that occurs in approximately 15% of multiple myeloma (MM) patients and confers a particularly poor prognosis. This study assesses the antimyeloma activity of
Tiina E Kähkönen et al.
Journal of bone oncology, 16, 100232-100232 (2019-04-09)
Advanced breast cancer has a high incidence of bone metastases. In bone, breast cancer cells induce osteolytic or mixed bone lesions by inducing an imbalance in bone formation and resorption. Activated fibroblast growth factor receptors (FGFRs) are important in regulation
Brian B Hasinoff et al.
Biochemical pharmacology, 84(12), 1617-1626 (2012-10-09)
Dovitinib (TKI258/CHIR258) is a multi-kinase inhibitor in phase III development for the treatment of several cancers. Dovitinib is a benzimidazole-quinolinone compound that structurally resembles the bisbenzimidazole minor groove binding dye Hoechst 33258. Dovitinib bound to DNA as shown by its
Daniel E Lopes de Menezes et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 11(14), 5281-5291 (2005-07-22)
Fms-like tyrosine kinase 3 (FLT3) encodes a receptor tyrosine kinase (RTK) for which activating mutations have been identified in a proportion of acute myelogenous leukemia (AML) patients and associated with poor clinical prognosis. Given the relevance of FLT3 mutations in
Yi-Han Chiu et al.
Journal of oncology, 2019, 2024648-2024648 (2019-09-06)
Breast cancer is the most common cancer and the leading cause of cancer deaths in women worldwide. The rising incidence rate and female mortality make it a significant public health concern in recent years. Dovitinib is a novel multitarget receptor

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service