Skip to Content
Merck
All Photos(1)

Key Documents

SML2931

Sigma-Aldrich

GSK264220A

≥98% (HPLC)

Synonym(s):

1-[[5-Methyl-4-[[(phenylamino)carbonyl]amino]-2-furanyl]sulfonyl]piperidine, GSK 264220A, N-[2-Methyl-5-(1-piperidinylsulfonyl)-3-furanyl]-N′-phenylurea

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H21N3O4S
CAS Number:
Molecular Weight:
363.43
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

[S](=O)(=O)(N3CCCCC3)c1[o]c(c(c1)NC(=O)Nc2ccccc2)C

InChI

1S/C17H21N3O4S/c1-13-15(19-17(21)18-14-8-4-2-5-9-14)12-16(24-13)25(22,23)20-10-6-3-7-11-20/h2,4-5,8-9,12H,3,6-7,10-11H2,1H3,(H2,18,19,21)

InChI key

LVOVQRPAMXCXTM-UHFFFAOYSA-N

Biochem/physiol Actions

GSK264220A is a potent, active site-targeting, covalent inhibitor against endothelial lipase (EL or LIPG; IC50 = 130 nM) and lipoprotein lipase (LPL or LIPD; IC50 = 100 nM). GSK264220A reduces NeuT expression - or CoCl2 treatment-induced intracellular triacylglycerol & lipid droplets (TAG & LD) increase in MCF-7 (16 & 32 nM) and suppresses LPL-depenent melanoma survival (by ~50% at 18.75 μM; WM852, WM1361). When applied in mice in vivo (30 mg/kg ip.), GSK264220A21 enhances lipid nanoparticles-mediated siRNAs delivery & gene-silencing efficacy.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cristina Cadenas et al.
International journal of cancer, 145(4), 901-915 (2019-01-18)
Endothelial lipase (LIPG) is a cell surface associated lipase that displays phospholipase A1 activity towards phosphatidylcholine present in high-density lipoproteins (HDL). LIPG was recently reported to be expressed in breast cancer and to support proliferation, tumourigenicity and metastasis. Here we
Yusuke Sato et al.
Molecular therapy : the journal of the American Society of Gene Therapy, 24(4), 788-795 (2015-12-19)
While a variety of short interfering RNA (siRNA) delivery compounds have been developed, a deep understanding of the key parameters that determine the quality of siRNA delivery are not known with certainty. Therefore, an understanding of the factors required for
Leslie E Lupien et al.
The Journal of pharmacology and experimental therapeutics, 371(1), 171-185 (2019-07-14)
It is well recognized that many cancers are addicted to a constant supply of fatty acids (FAs) and exhibit brisk de novo FA synthesis. Upregulation of a key lipogenic enzyme, fatty acid synthase (FASN), is a near-universal feature of human
Paul M Keller et al.
Journal of biomolecular screening, 13(6), 468-475 (2008-06-21)
Endothelial lipase (EL) is a 482-amino-acid protein from the triglyceride lipase gene family that uses a Ser-His-Asp triad for catalysis. Its expression in endothelial cells and preference for phospholipids rather than triglycerides are unique. Animal models in which it is
Krista B Goodman et al.
Bioorganic & medicinal chemistry letters, 19(1), 27-30 (2008-12-09)
Endothelial lipase (EL) activity has been implicated in HDL catabolism, vascular inflammation, and atherogenesis, and inhibitors are therefore expected to be useful for the treatment of cardiovascular disease. Sulfonylfuran urea 1 was identified in a high-throughput screening campaign as a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service