Skip to Content
Merck
All Photos(1)

Documents

SML2923

Sigma-Aldrich

ARV-771

≥98% (HPLC)

Synonym(s):

JQ1-VHL

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C49H60ClN9O7S2
CAS Number:
Molecular Weight:
986.64
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

ligand

VH032

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

Biochem/physiol Actions

ARV-771 is a bromodomain and extraterminal (BET) proteins degrader with a HIF-1?-derived von Hippel–Landau (VHL) E3 ligase-binding hydroxyproline and a BET-binding triazolo-diazepine acetamide. ARV-771 induces BET proteins degradation in castrate-resistent prostate cancer (CRPC) cultures (BRD2/3/4 DC50 <5 nM; 22Rv1, VCaP & LnCaP95) and reduces downstream c-Myc transcription with 10-500-fold higher potency than JQ-1, OTX015, and dBET1. ARV-771, but not JQ-1 or OTX015, effectively downregulates CRPC androgen receptor (30-300 nM) and causes CRPC tumor growth retardation/regression in mice in vivo (30 mg/kg s.c.; 22Rv1 and VCaP).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jonathan M Cooper et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 25(11), 3404-3416 (2019-02-24)
BET bromodomain inhibitors have emerged as a promising therapy for numerous cancer types in preclinical studies, including neurofibromatosis type 1 (NF1)-associated malignant peripheral nerve sheath tumor (MPNST). However, potential mechanisms underlying resistance to these inhibitors in different cancers are not
Neeraj Jain et al.
Science translational medicine, 11(497) (2019-06-21)
The activated B cell (ABC-like) subtype of diffuse large B cell lymphoma (DLBCL) is characterized by chronic activation of signaling initiated by immunoglobulin μ (IgM). By analyzing the DNA copy number profiles of 1000 DLBCL tumors, we identified gains of
Lu Zhang et al.
Molecular cancer therapeutics, 18(7), 1302-1311 (2019-05-09)
Proteolysis-targeting chimeras (PROTAC) are bifunctional molecules that hijack endogenous E3 ubiquitin ligases to induce ubiquitination and subsequent degradation of protein of interest. Recently, it has been shown that PROTACs with robust in vitro and in vivo activities and, in some
Philipp Ottis et al.
ACS chemical biology, 14(10), 2215-2223 (2019-09-26)
Proteolysis targeting chimeras are bifunctional small molecules capable of recruiting a target protein of interest to an E3 ubiquitin ligase that facilitates target ubiquitination followed by proteasome-mediated degradation. The first molecules acting on this novel therapeutic paradigm have just entered
Kanak Raina et al.
Proceedings of the National Academy of Sciences of the United States of America, 113(26), 7124-7129 (2016-06-09)
Prostate cancer has the second highest incidence among cancers in men worldwide and is the second leading cause of cancer deaths of men in the United States. Although androgen deprivation can initially lead to remission, the disease often progresses to

Related Content

Explore protein pathway analysis through chemical library screening, investigating protein interactions, and modulating signaling pathways with small molecules.

Explore protein pathway analysis through chemical library screening, investigating protein interactions, and modulating signaling pathways with small molecules.

Explore protein pathway analysis through chemical library screening, investigating protein interactions, and modulating signaling pathways with small molecules.

Explore protein pathway analysis through chemical library screening, investigating protein interactions, and modulating signaling pathways with small molecules.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service