Skip to Content
Merck
All Photos(1)

Documents

SML2747

Sigma-Aldrich

Forsythiaside A

≥98% (HPLC)

Synonym(s):

Forsythoside A, 2-(3,4-Dihydroxyphenyl)ethyl 6-O-(6-deoxy-α-L-mannopyranosyl)-4-[(2E)-3-(3,4-dihydroxyphenyl)-2-propenoate]-β-D-glucopyranoside, Forsythiaside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C29H36O15
CAS Number:
Molecular Weight:
624.59
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

optical activity

[α]/D -25 to -17°, c = 1 in methanol

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

InChI

1S/C29H36O15/c1-13-22(35)23(36)25(38)29(42-13)41-12-20-27(44-21(34)7-4-14-2-5-16(30)18(32)10-14)24(37)26(39)28(43-20)40-9-8-15-3-6-17(31)19(33)11-15/h2-7,10-11,13,20,22-33,35-39H,8-9,12H2,1H3/b7-4+/t13-,20+,22-,23+,24+,25+,26+,27+,28+,29+/m0/s1

InChI key

DTOUWTJYUCZJQD-UJERWXFOSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Forsythiaside A is a most abundant phenylethanoid glycosides isolated from Forsythia suspense that exhibits anti-inflammatory and neuroprotective activities. It appears that forsythoside A exerts antipyretic activities through inhibition of transient receptor potential vanilloid 1 (TRPV1) channel.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cuiling Liu et al.
International journal of biological sciences, 13(1), 65-75 (2017-01-27)
Transient receptor potential vanilloid 1 (TRPV1) is a non-selective cation channel gated by noxious heat, playing major roles in thermoregulation. Forsythoside A (FT-A) is the most abundant phenylethanoid glycosides in Fructus Forsythiae, which has been prescribed as a medicinal herb
Quan-Jiang Song et al.
PloS one, 11(9), e0162791-e0162791 (2016-09-13)
Bovine viral diarrhea virus (BVDV), the causative agent of bovine viral diarrhea/mucosal disease (BVD/MD), is an important pathogen of cattle and other wild animals throughout the world. BVDV infection typically leads to an impaired immune response in cattle. In the
Jin Qian et al.
European journal of pharmacology, 812, 250-255 (2017-07-25)
Forsythiaside A (FSA), an active constituent isolated from the Chinese medicinal herb Forsythia suspensa, has been known to have anti-inflammatory effect. However, the effect of FSA on allergic airway inflammation remains unclear. The aim of this study was to investigate

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service