Skip to Content
Merck
All Photos(1)

Key Documents

SML2692

Sigma-Aldrich

DCPIB

≥98% (HPLC), powder, VRAC or VSOAC blocker

Synonym(s):

4-(2-Butyl-6,7-dichloro-2-cyclopentyl-1-oxo-2,3-dihydro-1H-inden-5-yloxy)butanoic acid, 4-[(2-Butyl-6,7-dichloro-2-cyclopentyl-2,3-dihydro-1-oxo-1H-inden-5-yl)oxy]butanoic acid, 4-(2-butyl-6,7-dichloro-2-cyclopentylindan-1-on-5-yl)oxybutyric acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H28Cl2O4
CAS Number:
Molecular Weight:
427.36
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

product name

DCPIB, ≥98% (HPLC)

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

CCCCC1(Cc2cc(OCCCC(O)=O)c(Cl)c(Cl)c2C1=O)C3CCCC3

InChI

1S/C22H28Cl2O4/c1-2-3-10-22(15-7-4-5-8-15)13-14-12-16(28-11-6-9-17(25)26)19(23)20(24)18(14)21(22)27/h12,15H,2-11,13H2,1H3,(H,25,26)

InChI key

KHKGTPJPBOQECW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

DCPIB is a potent inhibitor of the volume-sensitive anion channel (VSAC). IC50 = 4.1μM ) versus ICl,swell in calf pulmonary artery endothelial (CPAE) cells. DCPIB does not inhibit CPAE cell ICl,Ca; hCFTR; nor guinea pig ICl,PKA, IKr, IKs,IKl, INa and ICa. DCPIB inhibition is reversible and voltage independent. ICl,swell is believed to be important in cardiovascular function. Studies have been limited by the lack of specific ligands.
DCPIB is a reversible and highly selective volume-regulated anion channel (VRAC or VSOAC) blocker (by 50/98.7% of hypotonic solution-induced Cl current or ICl,swell at 4.1/10 μM in calf pulmonary artery endothelial (CPAE) cells vs. 4.1% inhibition of Ca2+-activated ICl,Ca or CaCC at 10 μM) with little or no inhibition against hminK (IKs), hCFTR, hCLC-1/2/4/5, rCLC-K1, hKv1.5, hKv4.3, HERG (≤5.9% at 10 μM; transpected xenopus oocytes), nor IKs, IKr, IK1, INa, ICa, or ICl,PKA in isolated guinea pig cardiomyocytes. When applied in rats in vivo (10 mg/kg ip.), DCPIB is shown to protect against myocardial ischaemia/reperfusion injury.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yuesheng Xia et al.
Oncotarget, 7(26), 39345-39362 (2016-10-26)
Excessive reactive oxygen species (ROS) plays an important role in myocardial ischemia/reperfusion (I/R) injury, which triggers not only myocardial cellular apoptosis but also autophagy-related cell death, in which volume-sensitive outwardly rectifying (VSOR) Cl- channel-activated by ROS contributes to cell apoptotic
N Decher et al.
British journal of pharmacology, 134(7), 1467-1479 (2001-11-29)
1. We identified the ethacrynic-acid derivative DCPIB as a potent inhibitor of I(Cl,swell), which blocks native I(Cl,swell) of calf bovine pulmonary artery endothelial (CPAE) cells with an IC(50) of 4.1 microM. Similarly, 10 microM DCPIB almost completely inhibited the swelling-induced
David M Kern et al.
eLife, 8 (2019-02-19)
Hypoosmotic conditions activate volume-regulated anion channels in vertebrate cells. These channels are formed by leucine-rich repeat-containing protein 8 (LRRC8) family members and contain LRRC8A in homo- or hetero-hexameric assemblies. Here, we present single-particle cryo-electron microscopy structures of Mus musculus LRRC8A
Tingting Zhang et al.
Islets, 11(4), 77-88 (2019-03-09)
A potentiating effect of medium-chain triglycerides on glucose-stimulated insulin secretion (GSIS) has been observed since the 1960s. Subsequent observations identified octanoic acid (OA), the main component of medium-chain triglyceride, as the potentiator of GSIS, but the mechanism was unclear. We
Arturo Ponce et al.
Physiological reports, 7(11), e14029-e14029 (2019-06-13)
Giardia lamblia is one of the most important worldwide causes of intestinal infections, yet little is known about its cellular physiology, especially the diversity of ionic channels that this parasite expresses. In this work, we show that injection of mRNA

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service