Skip to Content
Merck
All Photos(1)

Key Documents

SML2390

Sigma-Aldrich

JNS 1-40

≥95% (HPLC)

Synonym(s):

N-benzyl-2-chloro-N-(2,3-dihydrobenzo[b][1,4]dioxin-6-yl)acetamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H16ClNO3
CAS Number:
Molecular Weight:
317.77
UNSPSC Code:
12352200

Assay

≥95% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C(CCl)N(CC1=CC=CC=C1)C2=CC3=C(OCCO3)C=C2

InChI

1S/C17H16ClNO3/c18-11-17(20)19(12-13-4-2-1-3-5-13)14-6-7-15-16(10-14)22-9-8-21-15/h1-7,10H,8-9,11-12H2

Biochem/physiol Actions

JNS 1-40 is a potent and selective cysteine-reactive covalent ligand of regulatory subunit PPP2R1A of the tumor-suppressor protein phosphatase 2A (PP2A) that activate PP2A in vitro and in vivo. JNS 1-40 targets C377 of PPP2R1A and inhibits breast cancer proliferation, and in vivo tumor growth.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Elizabeth A Grossman et al.
Cell chemical biology, 24(11), 1368-1376 (2017-09-19)
Many natural products that show therapeutic activities are often difficult to synthesize or isolate and have unknown targets, hindering their development as drugs. Identifying druggable hotspots targeted by covalently acting anti-cancer natural products can enable pharmacological interrogation of these sites

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service