Skip to Content
Merck
All Photos(1)

Key Documents

SML2335

Sigma-Aldrich

IACS-9571 trifluoroacetate salt

≥98% (HPLC)

Synonym(s):

IACS 009571-001-4 trifluoroacetate, N-(6-(3-(4-(Dimethylamino)butoxy)-5-propoxyphenoxy)-1,3-dimethyl-2-oxo-2,3-dihydro-1H-benzo[d]imidazol-5-yl)-3,4-dimethoxybenzenesulfonamide trifluoroacetate, N-[6-[3-[4-(Dimethylamino)butoxy]-5-propoxyphenoxy]-2,3-dihydro-1,3-dimethyl-2-oxo-1H-benzimidazol-5-yl]-3,4-dimethoxybenzenesulfonamide trifluoroacetate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H42N4O8S · CF3CO2H
CAS Number:
Molecular Weight:
756.79
UNSPSC Code:
12352200

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=S(C1=CC=C(C(OC)=C1)OC)(NC2=C(C=C(C3=C2)N(C(N3C)=O)C)OC4=CC(OCCC)=CC(OCCCCN(C)C)=C4)=O.FC(F)(C(O)=O)F

InChI

1S/C32H42N4O8S.C2HF3O2/c1-8-14-42-22-16-23(43-15-10-9-13-34(2)3)18-24(17-22)44-30-21-28-27(35(4)32(37)36(28)5)20-26(30)33-45(38,39)25-11-12-29(40-6)31(19-25)41-7;3-2(4,5)1(6)7/h11-12,16-21,33H,8-10,13-15H2,1-7H3;(H,6,7)

InChI key

GAOCLDPTFLDJIP-UHFFFAOYSA-N

Biochem/physiol Actions

High affinity, potent and selective dual BRPF1 & TRIM24 bromodomain (BrD) inhibitor with good pharmacokinetics and oral availability in mice in vivo.
IACS-9571 is a dimethylamine analogue.
IACS-9571 is a high-affinity, potent TRIM24/BRPF1 bromodomain (BrD) inhibitor (Kd = 1.3/2.1 nM by DiscoveRx) with good selectivity over other BrDs (BRPF2/3 Kd = 12/27 nM, BAZ2B/TAF1 BrD2/BRD4 BrD1,2 Kd = 0.4/1.8/>10 μM by DiscoveRx; ≤63% binding inhibition of 25 other BrDs at 1 μM). IACS-9571 potently blocks H3K23Ac peptide from TRIM24 BrD binding (IC50 = 7.6 nM) and displaces ectopically expressed TRIM24 PHD-BrD from endogenous histone H3 in 2-hr 5 μM SAHA-stimulated HeLa cells (IC50 = 50 nM). When adiministered in mice, IACS-9571 exhibits good pharmacokinetics and oral availability in vivo (F = 29%, 10 mg/kg p.o.).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Structure-guided design of IACS-9571, a selective high-affinity dual TRIM24-BRPF1 bromodomain inhibitor
Palmer WS, et al.
Journal of medicinal chemistry, 59(4), 1440-1454 (2015)
Lara N Gechijian et al.
Nature chemical biology, 14(4), 405-412 (2018-03-07)
The addressable pocket of a protein is often not functionally relevant in disease. This is true for the multidomain, bromodomain-containing transcriptional regulator TRIM24. TRIM24 has been posited as a dependency in numerous cancers, yet potent and selective ligands for the
Wylie S Palmer et al.
Journal of medicinal chemistry, 59(4), 1440-1454 (2015-06-11)
The bromodomain containing proteins TRIM24 (tripartite motif containing protein 24) and BRPF1 (bromodomain and PHD finger containing protein 1) are involved in the epigenetic regulation of gene expression and have been implicated in human cancer. Overexpression of TRIM24 correlates with
Yanai Zhan et al.
Epigenetics & chromatin, 8, 37-37 (2015-09-24)
Proteins that 'read' the histone code are central elements in epigenetic control and bromodomains, which bind acetyl-lysine motifs, are increasingly recognized as potential mediators of disease states. Notably, the first BET bromodomain-based therapies have entered clinical trials and there is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service