Skip to Content
Merck
All Photos(1)

Key Documents

SML2106

Sigma-Aldrich

Roflupram

≥98% (HPLC)

Synonym(s):

1-[4-(Difluoromethoxy)-3-[(tetrahydro-3-furanyl)oxy]phenyl]-3-methyl-1-butanone, FFPM, ZL-n-91

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H20F2O4
CAS Number:
Molecular Weight:
314.32
UNSPSC Code:
51111800
NACRES:
NA.77

Assay

≥98% (HPLC)

form

oil

color

colorless to light brown

storage temp.

2-8°C

SMILES string

O=C(CC(C)C)C1=CC=C(OC(F)F)C(OC2CCOC2)=C1

InChI key

IXURVUHDDXFYDR-UHFFFAOYSA-N

Biochem/physiol Actions

Roflupram (FFPM) is a potent anti-inflammatory phosphodiesterase 4 (PDE4) inhibitor that was found to suppress inflammasome activation in microglial cells by enhancing autophagy. In microglial BV-2 cells roflupram enhanced the level of microtubule-associated protein 1 light chain 3 II (LC3-II) and decreased the autophagy receptor ubiquitin-binding protein p62 (SQSTM1), enhancing autophagy, reducing the activation of inflammasome and suppressing the production of IL-1β. Roflupram (FFPM) was found to reverse learning and memory deficits in APP/PS1 transgenic mice.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tingting You et al.
ACS chemical neuroscience, 8(11), 2381-2392 (2017-06-13)
Inhibition of phosphodiesterase 4 (PDE4) suppressed the inflammatory responses in the brain. However, the underlying mechanisms are poorly understood. Roflupram (ROF) is a novel PDE4 inhibitor. In the present study, we found that ROF enhanced the level of microtubule-associated protein
Shilong Zheng et al.
Journal of medicinal chemistry, 51(24), 7673-7688 (2008-12-04)
Phosphodiesterase 4 catalyzes the hydrolysis of cyclic AMP and is a target for the development of anti-inflammatory agents. We have designed and synthesized a series of phenyl alkyl ketones as PDE4 inhibitors. Among them, 13 compounds were identified as having
Haibiao Guo et al.
Neuropharmacology, 116, 260-269 (2017-01-10)
Thus far, phosphodiesterase-4 (PDE4) inhibitors have not been approved for application in Alzheimer's disease (AD) in a clinical setting due to severe side effects, such as nausea and vomiting. In this study, we investigated the effect of FFPM, a novel
Hui-Fang Tang et al.
International immunopharmacology, 10(4), 406-411 (2010-01-16)
In the present study, we investigated the effect of classic PDE4 inhibitor rolipram and novel PDE4 inhibitor ZL-n-91 on LPS-induced acute lung injury (ALI) in mice and its mechanism. ALI was induced in ICR mice by instilling intratracheally with LPS
Dan Li et al.
International immunopharmacology, 90, 107176-107176 (2020-11-28)
Roflupram (ROF) is a novel phosphodiesterase 4 inhibitor. We previously found that ROF suppressed the production of pro-inflammatory factors in microglial cells; however, the underlying mechanisms are largely unknown. The present study aimed to elucidate the underlying molecular mechanisms of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service