Skip to Content
Merck
All Photos(1)

Key Documents

SML1118

Sigma-Aldrich

Hepcidin-25 (human) trifluoroacetate salt

≥95% (HPLC)

Synonym(s):

DTHFPICIFCCGCCHRSKCGMCCKT trifluoroacetate salt, H-Asp-Thr-His-Phe-Pro-Ile-Cys-Ile-Phe-Cys-Cys-Gly-Cys-Cys-His-Arg-Ser-Lys-Cys-Gly-Met-Cys-Cys-Lys-Thr-OH trifluoroacetate salt, Hepc25 (human), LEAP-1 (Liver-Expressed Antimicrobial Peptide) (human) trifluoroacetate salt, PLTR (Putative Liver Tumor Regressor ) (human)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C113H170N34O31S9 · xC2HF3O2
CAS Number:
Molecular Weight:
2789.35 (free base basis)
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥95% (HPLC)

form

powder

color

white to off-white

shipped in

wet ice

storage temp.

−20°C

Amino Acid Sequence

H-Asp-Thr-His-Phe-Pro-Ile-Cys-Ile-Phe-Cys-Cys-Gly-Cys-Cys-His-Arg-Ser-Lys-Cys-Gly-Met-Cys-Cys-Lys-Thr-OH trifluoroacetate salt

Biochem/physiol Actions

Hepcidin is the master regulator of iron homeostasis with anti-inflammatory and antimicrobial activity.
Hepcidin is the master regulator of iron homeostasis. Hepcidin is found in liver, but also widely distributed in brain, where it may play a role in preventing iron-related neurodegeneration. Hepcidin has anti-inflammatory and antimicrobial activity.
Hepcidin-25 is the major isoform of hepcidin. hepcidin‐25 with reticulocyte hemoglobin content (CHr) in the hepcidin‐25 plot helps to differentiate DA can be differentiated iron‐deficient anemia (IDA) from the anemia of chronic disease (ACD) and the combined state of ACD/IDA. Mutation in the hepcidin gene is associated with the development of severe juvenile hemochromatosis.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Association of prohepcidin and hepcidin-25 with erythropoietin response and ferritin in hemodialysis patients.
Kato A, et al.
American Journal of Nephrology, 28(1), 115-121 (2008)
Kyung-Cho Cho et al.
Journal of analytical methods in chemistry, 2018, 9653747-9653747 (2018-07-18)
As advanced mass spectrometry- (MS-) based hepcidin analysis offers to overcome the limitations in analytical methods using antihepcidin, further improvement of MS detection sensitivity for the peptide may enhance the diagnostic value of the hepcidin for various iron-related disorders. Here
Mutant antimicrobial peptide hepcidin is associated with severe juvenile hemochromatosis.
Roetto A, et al.
Nature Genetics, 33(1), 21-21 (2003)
Serum hepcidin?25 may replace the ferritin index in the Thomas plot in assessing iron status in anemic patients.
Thomas C, et al.
International Journal of Laboratory Hematology, 33(2), 187-193 (2011)
Jin-Seon Yook et al.
Proceedings of the National Academy of Sciences of the United States of America, 118(40) (2021-10-02)
Iron is an essential biometal, but is toxic if it exists in excess. Therefore, iron content is tightly regulated at cellular and systemic levels to meet metabolic demands but to avoid toxicity. We have recently reported that adaptive thermogenesis, a

Articles

DISCOVER Bioactive Small Molecules for Nitric Oxide & Cell Stress Research

DISCOVER Bioactive Small Molecules for Nitric Oxide & Cell Stress Research

DISCOVER Bioactive Small Molecules for Nitric Oxide & Cell Stress Research

DISCOVER Bioactive Small Molecules for Nitric Oxide & Cell Stress Research

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service