Skip to Content
Merck
All Photos(1)

Key Documents

SML1052

Sigma-Aldrich

2-D08

≥98% (HPLC)

Synonym(s):

2′,3′,4′-trihydroxy-flavone, 2-(2,3,4-Trihydroxyphenyl)-4H-1-Benzopyran-4-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H10O5
CAS Number:
Molecular Weight:
270.24
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

2-8°C

SMILES string

[o]1c2c([c](cc1c3c(c(c(cc3)O)O)O)=O)cccc2

InChI

1S/C15H10O5/c16-10-6-5-9(14(18)15(10)19)13-7-11(17)8-3-1-2-4-12(8)20-13/h1-7,16,18-19H

InChI key

JJAXTFSPCLZPIW-UHFFFAOYSA-N

Application

2-D08 has been used as a small ubiquitin-like modifier (SUMO)ylation inhibitor.

Biochem/physiol Actions

2-D08 is a potent and cell-permeable inhibitor of sumoylation that block the transfer of SUMO (small ubiquitin-like modifier) from the E2 enzyme (Ubc9) thioester conjugate to the substrate.
2-D08 is a potent and cell-permeable inhibitor of sumoylation.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

MG132?induced progerin clearance is mediated by autophagy activation and splicing regulation.
Harhouri K, et al.
EMBO Molecular Medicine (2017)
RAS GTPases are modified by SUMOylation.
Choi B H, et al.
Oncotarget, 9(4), 4440-4440 (2018)
Byeong Hyeok Choi et al.
Oncotarget, 9(4), 4440-4450 (2018-02-13)
RAS proteins are GTPases that participate in multiple signal cascades, regulating crucial cellular processes including cell survival, proliferation, differentiation, and autophagy. Mutations or deregulated activities of RAS are frequently the driving force for oncogenic transformation and tumorigenesis. Given the important
Pan Zhou et al.
Biochemical and biophysical research communications, 513(4), 1063-1069 (2019-04-24)
Acute myeloid leukemia (AML) is a heterogeneous clonal hematopoietic malignancy with poor survival and frequent relapse. Recently, a posttranslational modification of proteins with small ubiquitin-like modifiers (SUMO) has been notably implicated in a wide spectrum of diseases, especially cancers. Ubc9
Karim Harhouri et al.
EMBO molecular medicine, 9(9), 1294-1313 (2017-07-05)
Hutchinson-Gilford progeria syndrome (HGPS) is a lethal premature and accelerated aging disease caused by a de novo point mutation in LMNA encoding A-type lamins. Progerin, a truncated and toxic prelamin A issued from aberrant splicing, accumulates in HGPS cells' nuclei

Articles

DISCOVER Bioactive Small Molecules for Nitric Oxide & Cell Stress Research

DISCOVER Bioactive Small Molecules for Nitric Oxide & Cell Stress Research

DISCOVER Bioactive Small Molecules for Nitric Oxide & Cell Stress Research

DISCOVER Bioactive Small Molecules for Nitric Oxide & Cell Stress Research

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service