Skip to Content
Merck
All Photos(1)

Key Documents

SML1051

Sigma-Aldrich

Bivalirudin trifluoroacetate salt

≥97% (HPLC)

Synonym(s):

D-Phenylalanyl-L-prolyl-L-arginyl-L-prolylglycylglycylglycylglycyl-L-aspariginylglycyl-L-α-aspartyl-L-phenylalanyl-L-α-glutamyl-L-α-glutamyl-L-isoleucyl-L-prolyl-L-α-glutamyl-L-α-glutamyl-L-tyrosyl-L-leucine trifluoroacetate, Hirulog trifluoroacetate salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C98H138N24O33 · xC2HF3O2
CAS Number:
Molecular Weight:
2180.29 (free base basis)
UNSPSC Code:
51111800
NACRES:
NA.77

Assay

≥97% (HPLC)

form

powder

color

white to off-white

shipped in

wet ice

storage temp.

−20°C

SMILES string

N5([C@@H](CCC5)C(=O)N[C@@H](CCC(=O)O)C(=O)N[C@@H](CCC(=O)O)C(=O)N[C@@H](Cc6ccc(cc6)O)C(=O)N[C@@H](CC(C)C)C(=O)O)C(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)CNC(=O)[C@@H](NC(=O)CNC(=O)CNC(=O)CNC(=O)CNC(=O)[C@H]2N(CCC2)C(=O)[C@@H](

InChI

1S/C98H138N24O33/c1-5-52(4)82(96(153)122-39-15-23-70(122)92(149)114-60(30-34-79(134)135)85(142)111-59(29-33-78(132)133)86(143)116-64(43-55-24-26-56(123)27-25-55)89(146)118-67(97(154)155)40-51(2)3)119-87(144)61(31-35-80(136)137)112-84(141)58(28-32-77(130)131)113-88(145)63(42-54-18-10-7-11-19-54)117-90(147)66(45-81(138)139)110-76(129)50-107-83(140)65(44-71(100)124)109-75(128)49-106-73(126)47-104-72(125)46-105-74(127)48-108-91(148)68-21-13-38-121(68)95(152)62(20-12-36-103-98(101)102)115-93(150)69-22-14-37-120(69)94(151)57(99)41-53-16-8-6-9-17-53/h6-11,16-19,24-27,51-52,57-70,82,123H,5,12-15,20-23,28-50,99H2,1-4H3,(H2,100,124)(H,104,125)(H,105,127)(H,106,126)(H,107,140)(H,108,148)(H,109,128)(H,110,129)(H,111,142)(H,112,141)(H,113,145)(H,114,149)(H,115,150)(H,116,143)(H,117,147)(H,118,146)(H,119,144)(H,130,131)(H,132,133)(H,134,135)(H,136,137)(H,138,139)(H,154,155)(H4,101,102,103)/t52-,57+,58-,59-,60-,61-,62-,63-,64-,65-,66-,67-,68-,69-,70-,82-/m0/s1

InChI key

OIRCOABEOLEUMC-GEJPAHFPSA-N

Biochem/physiol Actions

Bivalirudin is a specific and reversible bivalent direct thrombin inhibitor.
Bivalirudin is a specific and reversible bivalent direct thrombin inhibitor. Bivalirudin specifically binds to both the catalytic site and to the anion-binding exosite of circulating and clot-bound thrombin.
Bivalirudin trifluoroacetate salt is a synthetic peptide composed of 20 amino acids. It serves as an anticoagulant for patients with unstable angina undergoing coronary angioplasty.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

John C Rohloff et al.
Molecular therapy. Nucleic acids, 3, e201-e201 (2014-10-08)
Limited chemical diversity of nucleic acid libraries has long been suspected to be a major constraining factor in the overall success of SELEX (Systematic Evolution of Ligands by EXponential enrichment). Despite this constraint, SELEX has enjoyed considerable success over the
Uzoma N Ibebuogu et al.
American journal of cardiovascular drugs : drugs, devices, and other interventions, 15(4), 275-285 (2015-03-19)
Diabetes mellitus (DM) is a pro-thrombotic state with enhanced thrombin generation and platelet reactivity. For most patients undergoing percutaneous coronary intervention (PCI), bivalirudin demonstrates efficacy comparable with that of heparin and glycoprotein IIb/IIIa inhibitors (GPIs). Yet, because of their pro-thrombotic
Matthew A Cavender et al.
Lancet (London, England), 384(9943), 599-606 (2014-08-19)
Bivalirudin is an alternative to heparin in patients undergoing percutaneous coronary intervention (PCI). We aimed to define the effects of a bivalirudin-based anticoagulation regimen compared with a heparin-based anticoagulation regimen on ischaemic and bleeding outcomes. We searched Medline, the Cochrane
Connie N Hess et al.
American heart journal, 170(2), 371-379 (2015-08-25)
In the SAFE-PCI for Women trial, patient preference for radial access for future procedures was greater than for femoral access. We sought to assess whether radial or femoral access impacts formal measures of quality-of-life (QOL) among women undergoing cardiac catheterization.
Matthew I Tomey et al.
American heart journal, 169(1), 86-93 (2014-12-17)
Women are more likely than men to experience adverse cardiac events after ST-elevation myocardial (STEMI). Whether differences in infarct size or reperfusion contribute to sex differences in outcomes is unknown. We compared baseline and procedural characteristics, angiographic and electrocardiographic indices

Articles

Bioactive small molecules for immune system signaling target identification/validation and antibiotics, antivirals, and antifungals offered.

Bioactive small molecules for immune system signaling target identification/validation and antibiotics, antivirals, and antifungals offered.

Bioactive small molecules for immune system signaling target identification/validation and antibiotics, antivirals, and antifungals offered.

Bioactive small molecules for immune system signaling target identification/validation and antibiotics, antivirals, and antifungals offered.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service