Skip to Content
Merck
All Photos(1)

Key Documents

SML0873

Sigma-Aldrich

Y16

≥98% (HPLC)

Synonym(s):

4-[[3-[(3-Methylphenyl)methoxy]phenyl]methylene]-1-phenyl-3,5-pyrazolidinedione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H20N2O3
CAS Number:
Molecular Weight:
384.43
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

, faint yellow to dark orange

solubility

DMSO: 5 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

N2(NC(=O)C(=Cc3cc(ccc3)OCc4cc(ccc4)C)C2=O)c1ccccc1

InChI

1S/C24H20N2O3/c1-17-7-5-9-19(13-17)16-29-21-12-6-8-18(14-21)15-22-23(27)25-26(24(22)28)20-10-3-2-4-11-20/h2-15H,16H2,1H3,(H,25,27)

InChI key

ITMLWGWTDWJSRZ-UHFFFAOYSA-N

Biochem/physiol Actions

Y16 blocks the binding of LARG, a DBL-family Rho guanine nucleotide exchange factor, with Rho (Kd = 80 nM). Y16 specifically inhibits LARG catalyzed activation of RhoA and RhoA signaling pathways. Y16 blocks the growth and migration of MCF7 breast cancer cells.

Features and Benefits

This compound is a featured product for Cyclic Nucleotide research. Click here to discover more featured Cyclic Nucleotide products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound is featured on the GTP Binding Proteins (Low Molecular Weight) page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Other Notes

Product is a racemic mixture of E/Z isomers.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sreeharsha Gurrapu et al.
Cell death and differentiation, 25(7), 1259-1275 (2018-03-21)
Semaphorin 4C (Sema4C) expression in human breast cancers correlates with poor disease outcome. Surprisingly, upon knock-down of Sema4C or its receptor PlexinB2 in diverse mammary carcinoma cells (but not their normal counterparts), we observed dramatic growth inhibition associated with impairment
Bo Zhang et al.
Cells, 9(3) (2020-03-08)
Rho GTPases, including Rho, Cdc42, Rac and ROP subfamilies, are key signaling molecules in RNA polymerase II (Pol II) transcriptional control. Our prior work has shown that plant ROP and yeast Cdc42 GTPases similarly modulate Ser2 and Ser5 phosphorylation status

Articles

Cyclic nucleotides like cAMP modulate cell function via PKA activation and ion channels.

Cyclic nucleotides like cAMP modulate cell function via PKA activation and ion channels.

Cyclic nucleotides like cAMP modulate cell function via PKA activation and ion channels.

Cyclic nucleotides like cAMP modulate cell function via PKA activation and ion channels.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service