Skip to Content
Merck
All Photos(1)

Key Documents

SML0847

Sigma-Aldrich

Benztropine mesylate

≥98% (HPLC)

Synonym(s):

Benzatropine mesylate, Benzotropine mesylate, Benztropine methanesulfonate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H25NO · CH3SO3H
CAS Number:
Molecular Weight:
403.53
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

H2O: 20 mg/mL, clear

storage temp.

2-8°C

SMILES string

CS(O)(=O)=O.CN1[C@H]2CC[C@@H]1C[C@@H](C2)OC(c3ccccc3)c4ccccc4

InChI

1S/C21H25NO.CH4O3S/c1-22-18-12-13-19(22)15-20(14-18)23-21(16-8-4-2-5-9-16)17-10-6-3-7-11-17;1-5(2,3)4/h2-11,18-21H,12-15H2,1H3;1H3,(H,2,3,4)/t18-,19+,20+;

InChI key

CPFJLLXFNPCTDW-BWSPSPBFSA-N

Gene Information

human ... CHRM1(1128)

Looking for similar products? Visit Product Comparison Guide

General description

Benztropine mesylate is an anti-histamine and dopamine re-uptake inhibitor.

Application

Benztropine mesylate has been used to study its target identification and mode of action against ebolavirus infection. It has also been used in pharmacological animal studies to evaluate its effect on SLC6A19 (solute carrier family 6 member 19; B0AT1).

Biochem/physiol Actions

Benztropine mesylate is a centrally acting muscarinic acetylcholine receptor antagonist and dopamine transporter (DAT) inhibitor (IC50 = 118 nM). Benztropine mesylate has been used to treat the symptoms of Parkinson′s disease and is currently in clincial trials for chronic back pain.
Benztropine mesylate serves as an inhibitor of breast cancer stem cells (BCSCs) in vitro and in vivo. It can help in improving the efficacy of chemotherapy in vitro. It is considered as an anti-cancer stem cell (CSC) drug, which can modify tumorigenic properties.

Features and Benefits

This compound is a featured product for Neuroscience research. Click here to discover more featured Neuroscience products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound is featured on the Acetylcholine Receptors (Muscarinic) and Biogenic Amine Transporters pages of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 4 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Target identification and mode of action of four chemically divergent drugs against ebolavirus infection
Ren J, et al.
Journal of medicinal chemistry, 61(3), 724-733 (2018)
Development of Biomarkers for Inhibition of SLC6A19 (B0AT1)?A Potential Target to Treat Metabolic Disorders
Javed K, et al.
Clinical Therapeutics, 19(11), 3597-3597 (2018)
New use of an old drug: inhibition of breast cancer stem cells by benztropine mesylate
Cui J, et al.
Oncotarget, 8(1), 1007-1007 (2017)
Björn Neumann et al.
Cell stem cell, 25(4), 473-485 (2019-10-05)
The age-related failure to produce oligodendrocytes from oligodendrocyte progenitor cells (OPCs) is associated with irreversible neurodegeneration in multiple sclerosis (MS). Consequently, regenerative approaches have significant potential for treating chronic demyelinating diseases. Here, we show that the differentiation potential of adult
Lisa M Johansen et al.
Science translational medicine, 7(290), 290ra89-290ra89 (2015-06-05)
Currently, no approved therapeutics exist to treat or prevent infections induced by Ebola viruses, and recent events have demonstrated an urgent need for rapid discovery of new treatments. Repurposing approved drugs for emerging infections remains a critical resource for potential

Articles

Muscarinic acetylcholine receptors mediate acetylcholine actions in CNS and non-nervous tissues, crucial for cell signaling.

Muscarinic acetylcholine receptors mediate acetylcholine actions in CNS and non-nervous tissues, crucial for cell signaling.

Muscarinic acetylcholine receptors mediate acetylcholine actions in CNS and non-nervous tissues, crucial for cell signaling.

Muscarinic acetylcholine receptors mediate acetylcholine actions in CNS and non-nervous tissues, crucial for cell signaling.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service