Skip to Content
Merck
All Photos(1)

Key Documents

SML0800

Sigma-Aldrich

Rimonabant hydrochloride

≥98% (HPLC), powder, cannabinoid type-I receptor antagonist

Synonym(s):

5-(4-Chlorophenyl)-1-(2,4-dichlorophenyl)-4-methyl-N-1-piperidinyl-1H-pyrazole-3-carboxamide hydrochloride, SR-141716, SR-141716A, SR141716, SR141716A

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H21Cl3N4O · HCl
CAS Number:
Molecular Weight:
500.25
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Product Name

Rimonabant hydrochloride, ≥98% (HPLC)

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

2-8°C

SMILES string

CC1=C(C2=CC=C(Cl)C=C2)N(C3=CC=C(Cl)C=C3Cl)N=C1C(NN4CCCCC4)=O.Cl

InChI

1S/C22H21Cl3N4O.ClH/c1-14-20(22(30)27-28-11-3-2-4-12-28)26-29(19-10-9-17(24)13-18(19)25)21(14)15-5-7-16(23)8-6-15;/h5-10,13H,2-4,11-12H2,1H3,(H,27,30);1H

InChI key

REOYOKXLUFHOBV-UHFFFAOYSA-N

Gene Information

human ... CNR1(1268)

Application

Rimonabant hydrochloride has been used as an antagonist of cannabinoid 1 (CB1) receptor:
  • to study its effects on protein synthesis in C2C12 myotubes
  • to analyze its effects on human astroglia
  • in combination with methanandamide (mAEA) to study its effects on murine gastric vagal afferent mechanosensitivity

Biochem/physiol Actions

Rimonabant acts as a mycobacterial membrane protein large 3 (MMPL3) inhibitor. Rimonabant hydrochloride exhibits therapeutic activity against weight reduction and smoking cessation.
Rimonabant hydrochloride (SR-141716A) is a potent and selective CB1 cannabinoid inverse agonist/antagonist with a Ki of 1.6 nM, minimal affinity for CB2, and and some GPR55 agonist activity. Rimonabant was developed as an anti-obesity drug because of its appetite suppressant activity, but was taken off the market because of side effects of depression and anxiety.
Rimonabant hydrochloride (SR-141716A) is a potent and selecvtive CB1 cannabinoid inverse agonist/antagonist with some GPR55 agonist activity and an appetite suppressant.

Features and Benefits

This compound is featured on the Cannabinoid Receptors and Cannabinoid Receptors pages of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A novel validated RP-HPLC method development for the estimation of rimonabant hydrochloride in bulk and tablet dosage forms
Nama S, et al.
African Journal of Pharmacy and Pharmacology, 5(2), 207-213 (2011)
Marc-Antoine Perrin et al.
Journal of pharmaceutical sciences, 102(7), 2311-2321 (2013-05-23)
Crystalline polymorphism occurs frequently in the solid state of active pharmaceutical ingredients, and this is problematic for the development of a suitable dose form. Rimonabant, an active pharmaceutical ingredient developed by Sanofi and discontinued because of side effects, exhibits dimorphism;
Jorge Emilio Ortega et al.
European journal of pharmacology, 709(1-3), 13-19 (2013-04-09)
Current pharmacological therapies for depression, including selective serotonin reuptake inhibitors (SSRI), are far from ideal. The cannabinoid system has been implicated in control of mood and neural processing of emotional information, and the modulation of serotonin (5-HT) release in the
Francisco Alen et al.
PloS one, 8(4), e60918-e60918 (2013-04-09)
Ghrelin is an endogenous regulator of energy homeostasis synthesized by the stomach to stimulate appetite and positive energy balance. Similarly, the endocannabinoid system is part of our internal machinery controlling food intake and energy expenditure. Both peripheral and central mechanisms
Alexandre Seillier et al.
Neuropsychopharmacology : official publication of the American College of Neuropsychopharmacology, 38(9), 1816-1824 (2013-04-09)
The neuronal mechanisms underlying social withdrawal, one of the core negative symptoms of schizophrenia, are not well understood. Recent studies suggest an involvement of the endocannabinoid system in the pathophysiology of schizophrenia and, in particular, of negative symptoms. We used

Articles

We offer many products related to cannabinoid receptors for your research needs.

We offer many products related to cannabinoid receptors for your research needs.

We offer many products related to cannabinoid receptors for your research needs.

We offer many products related to cannabinoid receptors for your research needs.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service