Skip to Content
Merck
All Photos(1)

Key Documents

SML0786

Sigma-Aldrich

Decursin

≥97% (HPLC)

Synonym(s):

(+)-Decursin, (7S)-7,8-Dihydro-8,8-dimethyl-2-oxo-2H,6H-benzo[1,2-b:5,4-b′]dipyran-7-yl 3-methyl-2-butenoate, 3-Methyl-2-butenoic acid (7S)-7,8-dihydro-8,8-dimethyl-2-oxo-2H,6H-pyrano[3,2-g]-1-benzopyran-7-yl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H20O5
CAS Number:
Molecular Weight:
328.36
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

biological source

Radix peucedani

Quality Level

Assay

≥97% (HPLC)

form

powder

optical activity

[α]/D +120 to +160°, c = 0.5 in chloroform-d

color

white to beige

solubility

DMSO: 5 mg/mL, clear

shipped in

wet ice

storage temp.

−20°C

SMILES string

[o]1c2c(cc3c(c2)OC([C@H](C3)OC(=O)C=C(C)C)(C)C)cc[c]1=O

InChI

1S/C19H20O5/c1-11(2)7-18(21)23-16-9-13-8-12-5-6-17(20)22-14(12)10-15(13)24-19(16,3)4/h5-8,10,16H,9H2,1-4H3/t16-/m0/s1

InChI key

CUKSFECWKQBVED-INIZCTEOSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Decursin is a cumarin isolated from Radix peucedani that exhibits a number of physiological actions including anticancer, antibacterial, antinematodal and antioxidant.
Decursin is a cumarin isolated from Radix peucedani that exhibits a number of physiological actions including anticancer, antibacterial, antinematodal and antioxidant. Decursin is potent anti-angiogenic agent targeting the VEGFR-2 signaling pathway. Decursin exhibits potent neuroprotective activity against glutamate and Ab- induced neurotoxicity. Apparently, decursin stimulates activation of Nerf2. Also, decursin inhibits androgen stimulated AR translocation to the nucleus in LNCaP prostate cancer cells.

Features and Benefits

This compound is featured on the Nuclear Receptors (Steroids) page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kyeong Lee et al.
European journal of medicinal chemistry, 45(12), 5567-5575 (2010-10-05)
We report the synthesis of a novel series of highly potent melanin inhibitors which were obtained through structural modification of an anticancer compound S-(+)-decursinol. The in vitro inhibitory potencies of the newly synthesized compounds were evaluated against α-MSH induced melanin
Seok Yun Jung et al.
Journal of cellular biochemistry, 113(5), 1478-1487 (2012-02-03)
Endothelial progenitor cells (EPCs) contribute to the tumor vasculature during tumor progression. Decursin isolated from the herb Angelica gigas is known to possess potent anti-inflammatory activities. Recently, we reported that decursin is a novel candidate for an angiogenesis inhibitor [Jung
Li Li et al.
Planta medica, 79(3-4), 275-280 (2013-02-01)
Decursin and decursinol angelate are the major components in the alcoholic extract of the root of Angelica gigas Nakai. Our previous work convincingly demonstrated that both decursin and decursinol angelate were rapidly converted to decursinol in mice after administration by
Jin Sook Song et al.
Xenobiotica; the fate of foreign compounds in biological systems, 41(10), 895-902 (2011-06-11)
Decursinol is a major coumarin derived from the roots of Angelica gigas and has various pharmacological effects against inflammation, angiogenesis, nociceptive pain and Alzheimer's disease. In vitro and in vivo studies were conducted to characterize the metabolism and pharmacokinetics of
Jung-woo Chae et al.
Journal of ethnopharmacology, 144(2), 248-254 (2012-09-18)
Decursin is used as a traditional Asian medicine to treat various women's diseases. Herb-drug interaction has become a serious problem since herbal medicine is extensively used in the modern world. This study investigates effects of decursin, on the pharmacokinetics of

Articles

We offers many products related to Nuclear Receptors (Steroids) for your research needs.

We offers many products related to Nuclear Receptors (Steroids) for your research needs.

We offers many products related to Nuclear Receptors (Steroids) for your research needs.

We offers many products related to Nuclear Receptors (Steroids) for your research needs.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service