SML0611
ML213
≥98% (HPLC)
Synonym(s):
N-(2,4,6-Trimethylphenyl)-bicyclo[2.2.1]heptane-2-carboxamide
About This Item
Recommended Products
Assay
≥98% (HPLC)
form
powder
color
white to beige
solubility
DMSO: 5 mg/mL, clear (warmed)
storage temp.
2-8°C
SMILES string
N(c3c(cc(cc3C)C)C)C(=O)C1C2CC(C1)CC2
InChI
1S/C17H23NO/c1-10-6-11(2)16(12(3)7-10)18-17(19)15-9-13-4-5-14(15)8-13/h6-7,13-15H,4-5,8-9H2,1-3H3,(H,18,19)
InChI key
SIQGKPGBLYKQBB-UHFFFAOYSA-N
Biochem/physiol Actions
Features and Benefits
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 4 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
We offer many products related to potassium channels for your research needs.
We offer many products related to potassium channels for your research needs.
We offer many products related to potassium channels for your research needs.
We offer many products related to potassium channels for your research needs.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service