Skip to Content
Merck
All Photos(1)

Key Documents

SML0440

Sigma-Aldrich

Spautin-1

≥98% (HPLC)

Synonym(s):

6-Fluoro-N-[(4-fluorophenyl)methyl]-4-quinazolinamine, C43

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H11F2N3
CAS Number:
Molecular Weight:
271.26
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 15 mg/mL (clear solution)

storage temp.

2-8°C

SMILES string

FC1=CC=C(C=C1)CNC2=C3C=C(F)C=CC3=NC=N2

InChI

1S/C15H11F2N3/c16-11-3-1-10(2-4-11)8-18-15-13-7-12(17)5-6-14(13)19-9-20-15/h1-7,9H,8H2,(H,18,19,20)

InChI key

AWIVHRPYFSSVOG-UHFFFAOYSA-N

Application

Spautin-1 has been used as an autophagy inhibitor:
  • to study its effects on vascular, glial, and neuronal alterations in the oxygen-induced retinopathy mouse model
  • to evaluate its pre-treatment effect on the response of canine osteosarcoma cells to doxorubicin
  • to study its effects on the production of interleukin (IL)-6 by oxidatively stressed dendritic cells (OS-DCs) in Luminex assay

Biochem/physiol Actions

Spautin-1 inhibits the activity of two ubiquitin-specific peptidases, USP10 and USP13, causing an increase in proteasomal degradation of class III PI3 kinase complexes, which have been shown to regulate autophagy.
Spautin-1 is an inhibitor of autophagy.
Specific and potent autophagy inhibitor-1 (Spautin-1) is a small molecule, which can inhibit autophagy and induce cancer cell death. It inhibits autophagy by degrading beclin-1. Spautin-1 augments the efficiency of radiation therapy and chemotherapy in various human cancer cell lines.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kuntal Ganguly et al.
Drug delivery, 23(8), 2838-2851 (2015-11-05)
Microspheres of chitosan (CS) cross-linked with polyethylene glycol (PEG) were prepared by emulsion-cross-linking followed by the solvent evaporation technique. The formulations were characterized and subjected to in vitro and in vivo tests to assess cell growth, changes in cell morphology
Dirk Mohn et al.
PloS one, 6(1), e16157-e16157 (2011-01-26)
Peri-implantitis has gained significant clinical attention in recent years. This disease is an inflammatory reaction to microorganisms around dental implants. Due to the limited accessibility, non-invasive antimicrobial strategies are of high interest. An unexpected approach to implant disinfection may evolve
Yanbin Ma et al.
PloS one, 8(2), e56500-e56500 (2013-02-26)
N-terminally his-tagged human mu opioid receptor, a G protein-coupled receptor was produced in E.coli employing synthetic codon-usage optimized constructs. The receptor was expressed in inclusion bodies and membrane-inserted in different E.coli strains. By optimizing the expression conditions the expression level
J P Adler-Moore et al.
Journal of liposome research, 27(3), 210-220 (2017-09-19)
Given the interest in the ectodomain of the matrix 2 (M2e) channel protein as a target for development of a universal influenza vaccine, we examined the role of the antigen configuration of M2e in generating a protective immune response. A
Ge Yang et al.
Scientific reports, 7(1), 17970-17970 (2017-12-23)
Presenilin (PS1 or PS2) functions as the catalytic subunit of γ-secretase, which produces the toxic amyloid beta peptides in Alzheimer's disease (AD). The dependence of folding and structural stability of PSs on the lipophilic environment and mutation were investigated by

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service