Skip to Content
Merck
All Photos(1)

Documents

SML0268

Sigma-Aldrich

AZD1152-HQPA

≥98% (HPLC)

Synonym(s):

3-[[7-[3-[Ethyl(2-hydroxyethyl)amino]propoxy]-4-quinazolinyl]amino]-N-(3-fluorophenyl)-1H-pyrazole-5-acetamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H30FN7O3
CAS Number:
Molecular Weight:
507.56
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: >15 mg/mL

storage temp.

2-8°C

SMILES string

CCN(CCO)CCCOc1ccc2c(Nc3cc(CC(=O)Nc4cccc(F)c4)[nH]n3)ncnc2c1

InChI

1S/C26H30FN7O3/c1-2-34(10-11-35)9-4-12-37-21-7-8-22-23(16-21)28-17-29-26(22)31-24-14-20(32-33-24)15-25(36)30-19-6-3-5-18(27)13-19/h3,5-8,13-14,16-17,35H,2,4,9-12,15H2,1H3,(H,30,36)(H2,28,29,31,32,33)

InChI key

QYZOGCMHVIGURT-UHFFFAOYSA-N

Application

AZD1152-HQPA has been used as a specific inhibitor of aurora kinase B (AURKB) to examine the cell cycle stage the IRG (hit compound that induces an irregular nuclear shape)-treated cells accumulate and become senescent and also to study the roles of cyclin-dependent kinase 6 (CDK6) and AURKB in restricting enterovirus 71 (EV71) infection.
AZD1152-HQPA has been used:
  • for the dose dependent inhibition of aurora kinase B (AURKB)
  • as a component to identify that, replication timing regulatory factor 1 (RIF1) and protein phosphatase 1 (PP1) are critical for the regulation of abscission timing in human cells
  • as a component to study the relevance of AURKB as a cancer therapeutic target

Biochem/physiol Actions

AZD1152-HQPA is the active metabolite of AZD-1152, a potent selective Aurora Kinase B inhibitor. AZD1152 is 50-fold selective for Aurora kinase B over Aurora kinase C and over 1000-fold selective for Aurora kinase B over Aurora Kinase A. (IC50s: aurora-A, 1,369 nmol/L; aurora-B, 0.36 nmol/L; aurora-C, 17.0 nmol/L). It is converted in plasma to the active form AZD1152-HQPA, which has been shown to have antineoplastic activity in a variety of animal models and human cancer cell lines.

Other Notes

AZD1152-HQPA has been expertly reviewed and recommended by the Chemical Probes Portal. For more information, please visit the AZD1152-HPQA probe summary on the Chemical Probes Portal website.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sandhya Noronha et al.
In vitro cellular & developmental biology. Animal, 54(1), 71-84 (2017-12-03)
Liposarcoma is a malignant soft tissue tumor that originates from adipose tissue and is one of the most frequently diagnosed soft tissue sarcomas in humans. There is great interest in identifying novel chemotherapeutic options for treating liposarcoma based upon molecular
Cell-based screen for altered nuclear phenotypes reveals senescence progression in polyploid cells after Aurora kinase B inhibition
Sadaie M, et al.
Molecular Biology of the Cell, 26(17), 2971-2985 (2015)
The RIF1-PP1 Axis Controls Abscission Timing in Human Cells
Bhowmick R, et al.
Current Biology (2019)
Human genome-wide RNAi screen reveals host factors required for enterovirus 71 replication
Wu KX, et al.
Nature Communications, 7, 13150-13150 (2016)
Ali Zekri et al.
Anti-cancer drugs, 28(8), 841-851 (2017-06-24)
Aurora-B kinase overexpression plays important roles in the malignant progression of prostate cancer (PCa). AZD1152-HQPA, as an inhibitor of Aurora-B, has recently emerged as a promising agent for cancer treatment. In this study, we aimed to investigate the effects of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service