Skip to Content
Merck
All Photos(1)

Key Documents

SML0224

Sigma-Aldrich

HI-6

≥98% (HPLC)

Synonym(s):

1-[[[4-(Aminocarbonyl)pyridinio]methoxy]methyl]-2-[(hydroxyimino)methyl]-pyridinium chloride, 4-Carbamoyl-1-[[[2-[(hydroxyimino)methyl]pyridinium-1-yl]methoxy]methyl]pyridinium dichloride, Asoxime chloride, HI 6, HI 6 chloride, HJ 6, Transant

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H16N4O3 · 2Cl
CAS Number:
Molecular Weight:
359.21
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

H2O: 15 mg/mL (clear solution)

shipped in

wet ice

storage temp.

−20°C

SMILES string

[Cl-].[Cl-].NC(=O)c1cc[n+](COC[n+]2ccccc2\C=N\O)cc1

InChI

1S/C14H14N4O3.2ClH/c15-14(19)12-4-7-17(8-5-12)10-21-11-18-6-2-1-3-13(18)9-16-20;;/h1-9H,10-11H2,(H-,15,19);2*1H

InChI key

QELSIJXWEROXOE-UHFFFAOYSA-N

General description

HI-6 functions as an antidote for treating intoxication caused by nerve agents. It acts as a potent drug in radiation protection and suppresses oxidative stress. HI-6 prevents the toxicity of the antineoplastic drug irinotecan.

Biochem/physiol Actions

HI-6 is an efficient oxime cholinesterase reactivator that is used as an antidote for organophosphates (Ops) exposure.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Modulation of ionising radiation generated oxidative stress by HI-6 (asoxime) in a laboratory rat model
Pohanka M, et al.
Neuro Endocrinology Letters, 31(2), 62-62 null
Miriam Dadparvar et al.
Toxicology letters, 206(1), 60-66 (2011-07-06)
The standard treatment of intoxication with organophosphorus (OP) compounds includes the administration of oximes acting as acetylcholinesterase (AChE) reactivating antidotes. However, the blood-brain barrier (BBB) restricts the rapid transport of these drugs from the blood into the brain in therapeutically
Jiri Kassa et al.
Basic & clinical pharmacology & toxicology, 110(4), 322-326 (2011-10-11)
The ability of three newly developed reversible inhibitors of acetylcholinesterase (AChE) (K298, K344 and K474) and currently available carbamate pyridostigmine to increase the resistance of mice against soman and the efficacy of antidotal treatment of soman-poisoned mice was compared. Neither
Reinhard Bogan et al.
Drug testing and analysis, 4(3-4), 208-214 (2012-02-24)
HI-6 exhibits superior efficacy in the therapy of intoxication by different highly toxic organophosphorus nerve agents. Therefore HI-6 is a promising candidate for the development of new antidotes against nerve agents. For ethical and safety reasons antidotes containing HI-6 should
Miroslav Pohanka et al.
Drug and chemical toxicology, 34(3), 255-260 (2011-06-09)
Oxime reactivator HI-6 (asoxime, in some sources) is a potent antidote suitable for treatment of intoxication by nerve agents. Despite the fact that HI-6 is considered for practical application in emergency situations, the impact of HI-6 on patients' bodies has

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service