Skip to Content
Merck
All Photos(1)

Key Documents

SML0198

Sigma-Aldrich

Fenoldopam mesylate

≥98% (HPLC)

Synonym(s):

Fenoldopam methanesulfonate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H16ClNO3 · CH4O3S
CAS Number:
Molecular Weight:
401.86
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to tan

mp

271 °C (lit.)

solubility

DMSO: ≥15 mg/mL at ~60 °C

storage temp.

room temp

SMILES string

CS(O)(=O)=O.Oc1ccc(cc1)C2CNCCc3c(Cl)c(O)c(O)cc23

InChI

1S/C16H16ClNO3.CH4O3S/c17-15-11-5-6-18-8-13(9-1-3-10(19)4-2-9)12(11)7-14(20)16(15)21;1-5(2,3)4/h1-4,7,13,18-21H,5-6,8H2;1H3,(H,2,3,4)

InChI key

CVKUMNRCIJMVAR-UHFFFAOYSA-N

Gene Information

human ... DRD1(1812)

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Fenoldopam mesylate may be used to study dopamine D1-mediated cell signaling.

Biochem/physiol Actions

Dopamine D1-receptor agonist; Antihypertensive
Fenoldopam mesylate (FM) is considered as an effective therapeutic agent to prevent the onset of postoperative AKI (PO-AKI). In healthy cats, FM can stimulate diuresis and be well-tolerated. This benzazepine-derivative is regional specific and does not cause cerebral vasodilation.
Fenoldopam mesylate is a selective dopamine D1 receptor agonist used as an antihypertensive agent. It is an agonist for D1-like dopamine receptors and binds with moderate affinity to α2-adrenoceptors. It has no significant affinity for D2-like receptors, α1 and α-adrenoceptors, 5HT1 and 5HT2 receptors, or muscarinic receptors. Fenoldopam mesylate causes peripheral vasodilation by stimulating dopamine-1receptors and α2-adrenoceptors, increasing renal blood flow.

Features and Benefits

This compound is featured on the Dopamine Receptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The pharmacokinetics of intravenous fenoldopam in healthy, awake cats
O'Neill KE, et al.
Journal of Veterinary Pharmacology and Therapeutics, 39, 202-204 (2016)
Pharmacokinetics and pharmacodynamics of fenoldopam mesylate for blood pressure control in pediatric patients
Hammer GB, et al.
BMC anesthesiology, 8(1), 6-6 (2008)
Fenoldopam Mesylate: A Narrative Review of Its Use in Acute Kidney Injury
Noce A, et al.
Current Pharmaceutical Biotechnology (2019)
51 What is the value of Non-Dialytic therapy in acute kidney injury
Evidence-Based Practice of Critical Care E-book: Expert Consult, 359-359 (2010)
A D Knapton et al.
Methods and findings in experimental and clinical pharmacology, 32(7), 489-497 (2010-11-12)
Exposure of the vasculature to vasodilators, pharmaceuticals and industrial chemicals may lead to injury of the blood vessel wall in animals. Vascular injury may begin with changes in the permeability of vascular endothelial cell and vessels, resulting in possible hemorrhage

Articles

We offer many products related to dopamine receptors for your research needs.

We offer many products related to dopamine receptors for your research needs.

We offer many products related to dopamine receptors for your research needs.

We offer many products related to dopamine receptors for your research needs.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service