Skip to Content
Merck
All Photos(1)

Key Documents

SML0119

Sigma-Aldrich

Acrivastine

≥98% (HPLC)

Synonym(s):

(E)-6-((E)-3-(1-Pyrrolidinyl)-1-p-tolylpropenyl)-2-pyridineacrylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H24N2O2
CAS Number:
Molecular Weight:
348.44
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: >2 mg/mL (warmed)

storage temp.

2-8°C

SMILES string

Cc1ccc(cc1)\C(=C/CN2CCCC2)c3cccc(\C=C\C(O)=O)n3

InChI

1S/C22H24N2O2/c1-17-7-9-18(10-8-17)20(13-16-24-14-2-3-15-24)21-6-4-5-19(23-21)11-12-22(25)26/h4-13H,2-3,14-16H2,1H3,(H,25,26)/b12-11+,20-13+

InChI key

PWACSDKDOHSSQD-IUTFFREVSA-N

Gene Information

human ... HRH1(3269)

Application

Acrivastine has been used as an antihistamine to investigate the relation between the increased residence time of antihistamine at the histamine H1 receptor (H1R) and the duration of effective target-inhibition by this antagonist.

Biochem/physiol Actions

Acrivastine is a second-generation H1-receptor antagonist.
Acrivastine is a second-generation antihistamine, an H1-receptor antagonist.
Acrivastine is a second-generation antihistamine. It is a derivative of first-generation compound triprolidine. Acrivastine is effectively used for treating allergic diseases including cholinergic urticaria and histamine-medicated dermatoses.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ilari Paakkari
Toxicology letters, 127(1-3), 279-284 (2002-06-08)
Although the new second-generation nonsedative antihistamines terfenadine and astemizole were launched as highly selective and specific H(1)-receptor antagonists, they were later found to cause prolongation of the QT-interval and severe cardiac arrhythmias. The prolongation of the QT-interval is caused by
F J de Abajo et al.
British journal of clinical pharmacology, 47(3), 307-313 (1999-04-24)
To quantify and compare the incidence of ventricular arrhythniias associated with the use of five nonsedating antihistamines: acrivastine, astemizole, cetirizine, loratadine and terfenadine. The effects of age, sex, dose, duration of treatment, and the interaction with P450 inhibitor drugs were
K Wieland et al.
The Journal of biological chemistry, 274(42), 29994-30000 (1999-10-09)
We combined in a previously derived three-dimensional model of the histamine H(1) receptor (Ter Laak, A. M., Timmerman, H., Leurs, H., Nederkoorn, P. H. J., Smit, M. J., and Donne-Op den Kelder, G. M. (1995) J. Comp. Aid. Mol. Design.
B K Ballmer-Weber et al.
Journal of investigational allergology & clinical immunology, 9(6), 351-355 (2000-02-09)
Certirizine, a potent H1-blocking agent, is often recommended as an emergency drug in anaphylactic reactions because of its well documented fast onset of action. In this randomized, cross-over study we compared the onset of action after a single dose of
R N Brogden et al.
Drugs, 41(6), 927-940 (1991-06-01)
Acrivastine is a short acting histamine H1-receptor antagonist with a rapid onset of action. Double-blind clinical trials have shown acrivastine (usually 8mg three times daily) to be an effective and well tolerated antihistamine in the treatment of chronic urticaria and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service