Skip to Content
Merck
All Photos(1)

Key Documents

SMB01027

Sigma-Aldrich

Pomiferin

≥90% (LC/MS-ELSD)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H24O6
CAS Number:
Molecular Weight:
420.45
MDL number:
UNSPSC Code:
12352205
NACRES:
NA.25

biological source

plant

Assay

≥90% (LC/MS-ELSD)

form

solid

mol wt

420.45

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

SMILES string

[o]1c2c([c](c(c1)c4cc(c(cc4)O)O)=O)c(c(c3c2C=CC(O3)(C)C)CC=C(C)C)O

InChI

1S/C25H24O6/c1-13(2)5-7-15-21(28)20-22(29)17(14-6-8-18(26)19(27)11-14)12-30-24(20)16-9-10-25(3,4)31-23(15)16/h5-6,8-12,26-28H,7H2,1-4H3

InChI key

GHCZYXUOYFOXIP-UHFFFAOYSA-N

General description

Pomiferin is a prenylated flavonoid and a bioactive natural compound commonly derived from plants such as Citrus aurantium, Derris montana, Maclura pomifera, and Acacia horrida. Current research indicates that this metabolite is an inhibitor and may possess diverse biological activities, including antimicrobial, anticancer, antioxidant, anti-inflammatory, and neuroprotective properties.

Application

Pomiferin is a natural product derived from plant source that finds application in compound screening libraries, metabolomics, phytochemical, and pharmaceutical research.

Biochem/physiol Actions

Pomiferin acts as an potential inhibitor of HDAC, with an IC50 of 1.05 μM, and also potently inhibits mTOR (IC50, 6.2 μM). Pomiferin shows cytotoxic effects on human tumor cell lines. Pomiferin also increases SOD activities and total antioxidative capacity, and decreases malondialdehyde in rats.

Features and Benefits

  • Suitable for Biochemical and Biomedical research
  • Versatile and adaptable for wide variety of laboratory and research applications

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Raymond Yang et al.
Journal of agricultural and food chemistry, 59(24), 13328-13336 (2011-11-18)
Pomiferin and osajin are prenylated isoflavones from Osage orange fruit that both have potent antioxidant activity in a variety of assays. Pomiferin, in particular, has strong activity against the superoxide anion in a photochemiluminescence (PCL) assay system. In vitro, pomiferin
Il Hong Son et al.
Bioorganic & medicinal chemistry letters, 17(17), 4753-4755 (2007-07-31)
The major constituents from the fruits of Maclura pomifera are the prenylated isoflavones, osajin (1) and pomiferin (2). Their structures were elucidated using NMR spectroscopic techniques and mass spectrometric analysis. Compound 2 showed potential inhibitory activity in histone deacetylase (HDAC)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service