SMB00805
L-Gulonic acid lithium salt
≥95%
Synonym(s):
Xylosecarboxylic acid lithium, (2S,3S,4R,5S)-2,3,4,5,6-Pentahydroxyhexanoic acid lithium salt, Lithium L-gulonate
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Empirical Formula (Hill Notation):
C6H11O7Li
Molecular Weight:
202.09
UNSPSC Code:
12352106
Recommended Products
General description
L-gulonic acid lithium salt, also known as Lithium L-gulonate, serves as a precursor in the synthesis of L-ascorbic acid, with potential applications in biochemical and metabolomics research. L-gulonic acid is a sugar acid belonging to the class of organic compounds known as medium-chain hydroxy acids and derivatives, characterized by a side chain consisting of 6 to 12 carbon atoms. Within the cell, gulonic acid is primarily located in the endoplasmic reticulum.
Application
L-gulonic acid lithium salt can be utilized to investigate microbial metabolism, explore enzyme functional diversity in metabolic processes, characterize metabolic pathways, and understand carbon utilization, providing potential benefits in biotechnology, drug discovery, cell culture, and biomedical research.
Features and Benefits
Versatile and adaptable for a wide variety of research and metabolomics applications
Other Notes
For additional information on our range of Biochemicals, please complete this form.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service