Skip to Content
Merck
All Photos(1)

Key Documents

SMB00085

Sigma-Aldrich

Asperuloside

≥95% (LC/MS-ELSD)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H22O11
CAS Number:
Molecular Weight:
414.36
EC Number:
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.25

Assay

≥95% (LC/MS-ELSD)

form

solid

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

SMILES string

CC(=O)OCC1=C[C@@H]2OC(=O)C3=COC(O[C@H]4O[C@H](CO)[C@@H](O)[C@H](O)[C@H]4O)[C@H]1[C@H]23

InChI

1S/C18H22O11/c1-6(20)25-4-7-2-9-12-8(16(24)27-9)5-26-17(11(7)12)29-18-15(23)14(22)13(21)10(3-19)28-18/h2,5,9-15,17-19,21-23H,3-4H2,1H3/t9-,10+,11+,12-,13+,14-,15+,17?,18+/m0/s1

InChI key

IBIPGYWNOBGEMH-ZORLMZJFSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Natural product derived from plant source.

Biochem/physiol Actions

Asperuloside has shown potential anti-obesity properties through its suppression of increased white adipose tissue, plasma triglycerides, and free fatty acids in the metabolic syndrome-like mouse model.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rosa Quirantes-Piné et al.
Journal of separation science, 33(17-18), 2818-2827 (2010-08-18)
In this study, a CE-MS method has been developed to carry out the qualitative characterization of a lemon verbena (Lippia citriodora) extract for the first time. The CE and ESI-MS parameters were optimized with respect to resolution, analysis time and
Zhitao Liang et al.
Phytomedicine : international journal of phytotherapy and phytopharmacology, 15(4), 259-267 (2008-03-15)
The herb of Oldenlandia diffusa (Willd.) Roxb. (Family Rubiaceae) is well-known in Chinese folk medicine for the treatment of hepatitis and malignant tumors of the liver, lung and stomach. However, another herb of O. corymbosa (L.) Lam is also used
Anti-obesity compounds in green leaves of Eucommia ulmoides
Hirata T, et al.
Bioorganic & Medicinal Chemistry Letters, 21(6), 1786-1791 (2011)
T Nakamura et al.
Mutation research, 388(1), 7-20 (1997-01-15)
The suppressing effect of crude extracts of Tochu tea, an aqueous extract of Eucommia ulmoides leaves and a popular beverage in Japan, on the induction of chromosome aberrations in CHO cells and mice was studied. When CHO cells were treated
Asperuloside monohydrate.
A Linden et al.
Acta crystallographica. Section C, Crystal structure communications, 56 ( Pt 5), 616-618 (2000-06-14)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service