Skip to Content
Merck
All Photos(1)

Key Documents

SHCLND

Sigma-Aldrich

MISSION® shRNA

Purified DNA

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106609

shipped in

dry ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

Our comprehensive shRNA product offering consists of over 150,000 pre-cloned shRNA constructs targeting more than 15,000 human and 15,000 mouse genes. The library is available in: gene family sets: gene collections related to specific functional classes such as kinases and ion channels; gene taget sets, sets of shRNA clones that target your favorite gene; and individual shRNA clones. Our shRNA and lentiviral manufacturing platforms utilize dedicated laboratories featuring state-of-the-art robotics, liquid handling and laboratory information management systems (LIMS) to provide superior quality and service for your RNAi research needs.
MISSION® TRC shRNA purified plasmid DNA format takes the hassle out of preparing DNA from each construct. DNA may be used directly for transient or stable transfection. Additionally these plasmids can be combined with packaging plasmids in a packaging cell line to produce lentiviral particles. 1 μg of DNA is provided for each construct in the target set.

Application

MISSION® shRNA has been used:
  • in the generation of ZO-1 (zonula occludens-1) knockdown
  • to knock down LIF (leukemia inhibitory factor) expression in isogenic MCF10 model
  • to knock down nuclear factor erythroid 2 like 1, in n MIN6 pancreatic β-cells

Other Notes

To see protocols and application data please visit sigma.com/shrna.

Legal Information

Use of this product is subject to one or more license agreements.
MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Deficiency of long isoforms of Nfe2l1 sensitizes MIN6 pancreatic beta cells to arsenite-induced cytotoxicity
Cui Q, et al.
Toxicology and Applied Pharmacology, 329(2), 67-74 (2017)
Epigenetic up-regulation of leukemia inhibitory factor (LIF) gene during the progression to breast cancer
Shin JE, et al.
Molecules and Cells, 31(2), 181-189 (2011)
Eva Ramos-Fernández et al.
Scientific reports, 11(1), 7395-7395 (2021-04-03)
Wnt signaling plays a key role in neurodevelopment and neuronal maturation. Specifically, Wnt5a stimulates postsynaptic assemblies, increases glutamatergic neurotransmission and, through calcium signaling, generates nitric oxide (NO). Trying to unveil the molecular pathway triggering these postsynaptic effects, we found that
Stage-specific roles of FGF2 signaling in human neural development
Grabiec M, et al.
Stem Cell Research, 17(2), 330-341 (2016)
Andrew R Chin et al.
Journal of mammary gland biology and neoplasia, 23(3), 165-176 (2018-07-04)
Extracellular vesicles (EVs) are secreted by many cell types and are increasingly investigated for their role in human diseases including cancer. Here we focus on the secretion and potential physiological function of non-pathological EVs secreted by polarized normal mammary epithelial

Articles

MISSION shRNA reduce the expression of specific target genes by targeting the specific mRNA therefore reducing the corresponding protein expression.

MISSION shRNA reduce the expression of specific target genes by targeting the specific mRNA therefore reducing the corresponding protein expression.

MISSION shRNA reduce the expression of specific target genes by targeting the specific mRNA therefore reducing the corresponding protein expression.

MISSION shRNA reduce the expression of specific target genes by targeting the specific mRNA therefore reducing the corresponding protein expression.

See All

Related Content

Find shRNAs for Individual Genes. Over 150,000 pre-cloned shRNA constructs targeting more than 15,000 human and 15,000 mouse genes.

Find shRNAs for Individual Genes. Over 150,000 pre-cloned shRNA constructs targeting more than 15,000 human and 15,000 mouse genes.

Find shRNAs for Individual Genes. Over 150,000 pre-cloned shRNA constructs targeting more than 15,000 human and 15,000 mouse genes.

Find shRNAs for Individual Genes. Over 150,000 pre-cloned shRNA constructs targeting more than 15,000 human and 15,000 mouse genes.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service