SCP0061
Calcitonin Gene Related Peptide II, cyclic human
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
Assay
≥95% (HPLC)
form
lyophilized
composition
Peptide Content, ≥75%
storage condition
protect from light
storage temp.
−20°C
Amino Acid Sequence
Ala-Cys-Asn-Thr-Ala-Thr-Cys-Val-Thr-His-Arg-Leu-Ala-Gly-Leu-Leu-Ser-Arg-Ser-Gly-Gly-Met-Val-Lys-Ser-Asn-Phe-Val-Pro-Thr-Asn-Val-Gly-Ser-Lys-Ala-Phe-NH2
Application
Calcitonin Gene Related Peptide II (CGRP-II) is a member of the calcitonin gene family used to study CGRP receptor specificity and function and CGRP signaling.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Critical reviews in neurobiology, 11(2-3), 167-239 (1997-01-01)
The calcitonin gene peptide superfamily consists of calcitonin (CT), calcitonin gene-related peptide (CGRP), and amylin. CT and CGRP derive from the CT/CGRP gene, which is encoded on chromosome 11. Alternative splicing of the primary RNA transcript leads to the translation
Endocrinology, 146(6), 2699-2708 (2005-03-12)
Human adipose tissue is a contributor to inflammation- and sepsis-induced elevation of serum procalcitonin (ProCT). Several calcitonin (CT) peptides, including ProCT, CT gene-related peptide (CGRP), and adrenomedullin (ADM) are suspected mediators in human inflammatory diseases. Therefore, we aimed to explore
Endocrinology, 153(4), 1850-1860 (2012-02-09)
Calcitonin gene-related peptide (CGRP) is a neuropeptide with multiple neuroendocrine roles, including vasodilation, migraine, and pain. The receptor for CGRP is a G protein-coupled receptor (GPCR) that requires three proteins for function. CGRP binds to a heterodimer composed of the
Bone, 43(5), 813-818 (2008-08-09)
The 'calcitonin family' is a group of peptide hormones that share structural similarities with calcitonin, and includes calcitonin gene-related peptide (CGRP), amylin, adrenomedullin and adrenomedullin 2 (intermedin). These hormones are produced by different tissues, with calcitonin being produced in thyroid
Molecular and cellular endocrinology, 199(1-2), 119-128 (2003-02-13)
In this study, we describe the cloning and tissue expression of equine calcitonin (CT), calcitonin-gene related peptide (CGRP)-I, and CGRP-II cDNA. We also describe a novel divergent form of CGRP (CGRP-I). Equine CT has greatest homology (>85%) to human, rat
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service