Skip to Content
Merck
All Photos(2)

Key Documents

SBR00068

Sigma-Aldrich

Streptomycin Ready Made Solution

100 mg/mL in water

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
51281624
NACRES:
NA.76
Pricing and availability is not currently available.

form

liquid

Quality Level

specific activity

(Antibacterial)

concentration

100 mg/mL in water

antibiotic activity spectrum

Gram-negative bacteria
Gram-positive bacteria
mycobacteria

Mode of action

protein synthesis | interferes

storage temp.

−20°C

SMILES string

[S](=O)(=O)(O)O.[S](=O)(=O)(O)O.[S](=O)(=O)(O)O.N([C@@H]4[C@@H](O[C@H]([C@@H]([C@H]4O)O)CO)O[C@H]5[C@@H](O[C@H]([C@]5(O)C=O)C)O[C@H]6[C@@H]([C@H]([C@@H]([C@H]([C@@H]6O)O)N=C(N)N)O)N=C(N)N)C.N([C@@H]1[C@@H](O[C@H]([C@@H]([C@H]1O)O)CO)O[C@H]2[C@@H](O[C@H]([

InChI

1S/2C21H39N7O12.3H2O4S/c2*1-5-21(36,4-30)16(40-17-9(26-2)13(34)10(31)6(3-29)38-17)18(37-5)39-15-8(28-20(24)25)11(32)7(27-19(22)23)12(33)14(15)35;3*1-5(2,3)4/h2*4-18,26,29,31-36H,3H2,1-2H3,(H4,22,23,27)(H4,24,25,28);3*(H2,1,2,3,4)/t2*5-,6-,7-,8+,9-,10-,11-

InChI key

QTENRWWVYAAPBI-YZTFXSNBSA-N

General description

Chemical structure: aminoglycoside

Application

Streptomycin is used to study mechanisms of streptomycin resistance and is often used together with penicillin and other agents to inhibit bacterial contamination in cell culture applications. It is recommended for use in molecular biology applications at 25-50 μg/mL, in cell culture applications at 100 mg/L and in embryo culture at 50 mg/L.

Biochem/physiol Actions

Mode of Action: Streptomycin acts by inhibiting prokaryote protein synthesis by binding to the S12 protein of the 30S ribosomal subunit, thus preventing the transition from imitation complex to chain-elongating ribosome. This causes miscoding or inhibits initiation.

Mode of Resistance: A mutation in rpsL, a gene for S12 ribosomal protein, prevents binding of streptomycin to the ribosome. An aminoglycoside phosphotransferase also inactivates streptomycin.

Antimicrobial spectrum: Streptomycin acts against gram-negative and gram-positive bacteria.

Storage and Stability

  • Streptomycin ready made solution contains 100 mg/mL streptomycin sulfate in water.
  • Streptomycin: sulfate salt ratio is 1:1.5.
  • The product is packed under aseptic conditions.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Repr. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jiayun Zhu et al.
Cell cycle (Georgetown, Tex.), 14(12), 1809-1822 (2015-06-18)
Fanconi Anemia (FA) is a cancer predisposition syndrome and the factors defective in FA are involved in DNA replication, DNA damage repair and tumor suppression. Here, we show that FANCD2 is critical for genome stability maintenance in response to high-linear
Daniela Lobenwein et al.
Journal of the American Heart Association, 4(10), e002440-e002440 (2015-10-29)
Paraplegia following spinal cord ischemia represents a devastating complication of both aortic surgery and endovascular aortic repair. Shock wave treatment was shown to induce angiogenesis and regeneration in ischemic tissue by modulation of early inflammatory response via Toll-like receptor (TLR)
Rabia T Khan et al.
PloS one, 9(2), e88009-e88009 (2014-02-08)
Experimental animal models of Salmonella infections have been widely used to identify genes important in the host immune response to infection. Using an F2 cross between the classical inbred strain C57BL/6J and the wild derived strain MOLF/Ei, we have previously
Carlotta Alias et al.
Journal of cellular and molecular medicine, 19(9), 2098-2107 (2015-05-27)
Gynaecological leiomyosarcoma (gLMS) represent a heterogeneous group of soft tissue sarcoma, characterized by rare incidence, high aggressiveness and propensity to infiltrate secondary organs, poor prognosis and lethality, because of the lack of biological mechanisms that underlying their progression and effective
Manuel Lutzenberger et al.
PloS one, 10(7), e0134228-e0134228 (2015-08-01)
Alzheimer's disease (AD) and prion diseases carry a significant inflammatory component. The astrocytic overexpression of CCAAT/enhancer-binding protein delta (C/EBPD) in prion- and AD-affected brain tissue prompted us to study the role of this transcription factor in murine model systems of

Questions

  1. Streptomycin ready made solution 100mg/ml cat # SBR00068 : is the solution filtered at the end of the preparation? Thank you

    1 answer
    1. The details of the manufacturing processing cannot be shared, for proprietary reasons, but the product is not sterile filtered. It is is packaged under aseptic conditions.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service