SAE0181
3CL, Mpro protease chromogenic substrate peptide
pNA-Peptide substrate, ≥95% (HPLC), lyophilized
Synonym(s):
3CL protease peptide, COVID-19 detection, Thr-Ser-Ala-Val-Leu-Gln-pNA
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352202
NACRES:
NA.22
Recommended Products
Product Name
3CL, Mpro protease chromogenic substrate peptide, pNA-Peptide substrate, lyophilized
Quality Level
shipped in
ambient
storage temp.
−20°C
General description
This peptide is a substrate for the Main protease from SARS-Cov2, 3CL protease (Mpro), . This substrate allows measurement of the activity of 3CL protease using a standard plate reader or spectrophotometer.
Proteolytic cleavage by 3CL protease will release the free pNA group from the peptide (sequence TSAVLQ) with maximum absorption peak at 405 nm (A4051μM = 0.00916)
Peptide sequence: Thr-Ser-Ala-Val-Leu-Gln-pNA
Lyophilized from 0.1% TFA in H2O
Proteolytic cleavage by 3CL protease will release the free pNA group from the peptide (sequence TSAVLQ) with maximum absorption peak at 405 nm (A4051μM = 0.00916)
Peptide sequence: Thr-Ser-Ala-Val-Leu-Gln-pNA
Lyophilized from 0.1% TFA in H2O
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service