SAB4700662
Monoclonal Anti-HLA-DR-PE antibody produced in mouse
clone MEM-12, purified immunoglobulin, buffered aqueous solution
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352203
NACRES:
NA.41
Recommended Products
biological source
mouse
conjugate
phycoerythrin (R-PE) conjugate
antibody form
purified immunoglobulin
antibody product type
primary antibodies
clone
MEM-12, monoclonal
form
buffered aqueous solution
species reactivity
human
technique(s)
flow cytometry: suitable
isotype
IgG1
shipped in
wet ice
storage temp.
2-8°C
target post-translational modification
unmodified
Gene Information
human ... HLA-DRA(3122)
General description
Human leukocyte antigen (HLA)-DR is a member of HLA class II molecules. It is encoded by the gene mapped to human chromosome 6. HLA class II molecules are highly expressed on B-Iymphocytes, but is also found at minimal levels on T-Iymphocytes, macrophages, thymus epithelial cells and selected cells of various body tissues. HLA-DR consists of a non-polymorphic α chain gene and up to three distinct highly polymorphic β chain genes.
The antibody MEM-12 recognizes common epitope on human HLA-DR which is dependent on the association of alpha and beta chains. DR is the isotype of human MHC Class II molecules expressed on antigen-presenting cells (APC; dendritic cells, B lymphocytes, monocytes, macrophages).
Immunogen
Thymocyte membrane
Application
The reagent is designed for Flow Cytometry analysis of human blood cells using 20 μL reagent / 100 μL of whole blood or 1e6 cells in a suspension. The content of a vial (2 mL) is sufficient for 100 tests.
Biochem/physiol Actions
Human leukocyte antigen (HLA)-DR activates a T-cell receptor by presenting antigenic peptides derived from extracellular proteins thus, stimulating an immune response against the antigen. Mutation in the gene increases the risk of susceptibility to inflammatory bowel disease (IBD), Crohn′s disease and ulcerative colitis. Overexpression of monocytic (HLA)-DR has been observed in schizophrenic patients.
Features and Benefits
Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.
Physical form
Solution in phosphate buffered saline containing 15 mM sodium azide and 0.2% high-grade protease free BSA as a stabilizing agent.
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
10 - Combustible liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Monocytic HLA DR antigens in schizophrenic patients.
Krause D
Neuroscience Research, 72, 87-93 (2012)
HLA-DR and -DQ phenotypes in inflammatory bowel disease: a meta-analysis.
Stokkers PC
Gut, 45, 395-401 (1999)
Reactive microglia in patients with senile dementia
of the Alzheimer type are positive for the
histoconlpatibility glycoprotein HLA-DR
of the Alzheimer type are positive for the
histoconlpatibility glycoprotein HLA-DR
McGeer PL
Neuroscience Letters, 79, 195-200 (1987)
HLA-DR peptide occupancy can be regulated with a wide variety of small molecules.
Blazekovic F
Human Vaccines & Immunotherapeutics, 12, 593-598 (2016)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service