SAB4600036
Anti-Rabbit IgG (H+L), highly cross-adsorbed, CF™ 488A antibody produced in donkey
~2 mg/mL, affinity isolated antibody
Sign Into View Organizational & Contract Pricing
All Photos(6)
About This Item
UNSPSC Code:
12352203
NACRES:
NA.46
Recommended Products
biological source
donkey
Quality Level
conjugate
CF™ 488A conjugate
antibody form
affinity isolated antibody
antibody product type
secondary antibodies
clone
polyclonal
form
buffered aqueous solution
species reactivity
rabbit
concentration
~2 mg/mL
technique(s)
flow cytometry: 1-10 μg/mL
immunocytochemistry: suitable
immunohistochemistry: suitable
indirect immunofluorescence: 1-10 μg/mL
fluorescence
λex 490 nm; λem 515 nm
shipped in
ambient
storage temp.
−20°C
target post-translational modification
unmodified
General description
Immunoglobulin G (IgG) belongs to the immunoglobulin family and is a widely expressed serum antibody. An immunoglobulin has two heavy chains and two light chains connected by a disulfide bond. It is a glycoprotein and mainly helps in immune defense. IgG is a major class of immunoglobulin. Immunoglobulin G (IgG) participates in hypersensitivity type II and type III. Rabbit IgG is further divided into five classes- IgM, IgG, IgA, IgD and IgE. It has only one IgG variant with one interheavy disulfide bridge. It helps in opsonization, complement fixation and antibody dependent cell mediated cytotoxicity.
Specificity
Binds all rabbit IgGs (minimal cross-reaction with bovine, chicken, goat, guinea pig, hamster, horse, human, mouse, rat and sheep serum proteins)
Immunogen
rabbit IgG (H+L)
Application
Anti-Rabbit IgG (H+L), highly cross-adsorbed, CF™ 488A antibody has been used in immunocytochemistry.
Features and Benefits
Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.
Physical form
Supplied in phosphate buffered saline with 0.05% sodium azide, 50% glycerol and 2 mg/mL bovine serum albumin.
Preparation Note
Protect from light.
Legal Information
This product is distributed by Sigma-Aldrich Co. under the authorization of Biotium, Inc. This product is covered by one or more US patents and corresponding patent claims outside the US patents or pending applications owned or licensed by Biotium, Inc. including without limitation: 12/334,387; 12/607,915; 12/699,778; 12/850,578; 61/454,484. In consideration of the purchase price paid by the buyer, the buyer is hereby granted a limited, non-exclusive, non-transferable license to use only the purchased amount of the product solely for the buyer′s own internal research in a manner consistent with the accompanying product literature. Except as expressly granted herein, the sale of this product does not grant to or convey upon the buyer any license, expressly, by implication or estoppel, under any patent right or other intellectual property right of Biotium, Inc. Buyer shall not resell or transfer this product to any third party, or use the product for any commercial purposes, including without limitation, any diagnostic, therapeutic or prophylactic uses. This product is for research use only. Any other uses, including diagnostic uses, require a separate license from Biotium, Inc. For information on purchasing a license to use this product for purposes other than research, contact Biotium, Inc., 3159 Corporate Place, Hayward, CA 94545, Tel: (510) 265-1027. Fax: (510) 265-1352. Email: [email protected].
CF is a trademark of Biotium, Inc.
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
10 - Combustible liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Default patterning produces pan-cortical glutamatergic and CGE/LGE-like GABAergic neurons from human pluripotent stem cells.
Floruta CM, et al.
Stem Cell Reports, 9(5), 1463-1476 (2017)
Lymphoblastoid cell lines as a model to understand amyotrophic lateral sclerosis disease mechanisms.
Orietta Pansarasa et al.
Disease models & mechanisms, 11(3) (2018-02-09)
In the past, amyotrophic lateral sclerosis (ALS) has been considered a 'neurocentric' disease; however, new evidence suggests that it should instead be looked at from a 'multisystemic' or 'non-neurocentric' point of view. From 2006, we focused on the study of
Chellathurai Vasantha Niranjan et al.
Reports of biochemistry & molecular biology, 9(3), 264-269 (2021-03-03)
Studying protein-protein and protein-DNA interactions are prerequisites for the identification of function and mechanistic role of various proteins in the cell. Protocols for analyzing DNA-based Protein-Protein and Protein-DNA interactions are complicated and need to be simplified for efficient tracking of
Optimized synthesis and pharmacological evaluation of HCN channel inhibitor EC18.
Patberg, et al.
Arch. Pharm. (Weinheim), 356, e2200665-e2200665 (2023)
Matteo Bordoni et al.
Journal of clinical medicine, 8(5) (2019-05-28)
We already demonstrated that in peripheral blood mononuclear cells (PBMCs) of sporadic amyotrophic lateral sclerosis (sALS) patients, superoxide dismutase 1 (SOD1) was present in an aggregated form in the cytoplasmic compartment. Here, we investigated the possible effect of soluble SOD1
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service