Skip to Content
Merck
All Photos(12)

Key Documents

SAB4504388

Sigma-Aldrich

Anti-phospho-eIF2 α (pSer51) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-EIF-2, Anti-EIF-2A, Anti-EIF-2alpha, Anti-EIF2, Anti-EIF2A

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 36 kDa

species reactivity

human, mouse, rat

concentration

~1 mg/mL

technique(s)

ELISA: 1:10000
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

phosphorylation (pSer51)

Gene Information

human ... EIF2S1(1965)

General description

EIF2S1 (eukaryotic translation initiation factor 2 subunit α) is a heterotrimeric protein and a regulatory subunit of eIF2 . eIF2 has three general subunits- α, βand γ . eIF2α is located on human chromosome 14q23 .

Immunogen

The antiserum was produced against synthesized peptide derived from human eIF2 alpha around the phosphorylation site of Ser51.

Immunogen Range: 21-70

Application

Anti-phospho-eIF2 α(pSer51) antibody has been used in western blotting and immunohistochemistry.

Biochem/physiol Actions

EIF2S1 (eukaryotic translation initiation factor 2 subunit α) acts as a modulator in apoptosis signaling pathway . During endoplasmic reticulum stress, eIF2α plays a major role in signaling pathways in the development of tumor .

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Isabella R Straub et al.
Human molecular genetics, 30(8), 687-705 (2021-03-23)
Mutations in CHCHD10, coding for a mitochondrial intermembrane space protein, are a rare cause of autosomal dominant amyotrophic lateral sclerosis. Mutation-specific toxic gain of function or haploinsufficiency models have been proposed to explain pathogenicity. To decipher the metabolic dysfunction associated
The Role of the PERK/eIF2?/ATF4/CHOP Signaling Pathway in Tumor Progression During Endoplasmic Reticulum Stress
Rozpedek W, et al.
Current Molecular Medicine, 16(6), 533-544 (2016)
Tsz Hang Wong et al.
Neurobiology of aging, 73, 229-229 (2018-10-14)
Next-generation sequencing has contributed to our understanding of the genetics of Alzheimer's disease (AD) and has explained a substantial part of the missing heritability of familial AD. We sequenced 19 exomes from 8 Dutch families with a high AD burden
Translational regulator eIF2? in tumor
Zheng Q, et al.
Tumour Biology : the Journal of the International Society For Oncodevelopmental Biology and Medicine, 35(7), 6255-6264 (2014)
6-Shogaol induces apoptosis in human leukemia cells through a process involving caspase-mediated cleavage of eIF2?
Liu Q, et al.
Molecular Cancer, 12(1), 135-135 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service