Skip to Content
Merck
All Photos(2)

Key Documents

SAB4504126

Sigma-Aldrich

Anti-phospho-Caspase 3 (pSer150) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-CASP-3, Anti-Caspase-3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 31 kDa

species reactivity

mouse, rat, human

concentration

~1 mg/mL

technique(s)

ELISA: 1:40000
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

phosphorylation (pSer150)

Gene Information

human ... CASP3(836)

General description

Caspase-3 (CASP3) belongs to the class of cysteine proteases. CASP3 is an executioner caspase, which is cleaved and activated with the help of caspase-8 and caspase-9 initiator caspases. CASP3 is mapped to human chromosome 4q35.1.

Immunogen

The antiserum was produced against synthesized peptide derived from human Caspase 3 around the phosphorylation site of Ser150.

Immunogen Range: 116-165

Biochem/physiol Actions

Caspase-3 (CASP3) can degenerate multiple cellular proteins and results in morphological changes and DNA fragmentation in cells at the time of apoptosis. It participates in the growth stimulation. CASP3 plays a major role in the execution phase of apoptosis.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Expression levels of cleaved caspase-3 and caspase-3 in tumorigenesis and prognosis of oral tongue squamous cell carcinoma.
Liu PF, et al.
PLoS ONE, 12(7), e0180620-e0180620 (2017)
Caspase 3?mediated stimulation of tumor cell repopulation during cancer radiotherapy.
Huang Q, et al.
Nature Medicine, 17(7), 860?866-860?866 (2011)
CASP3 gene single-nucleotide polymorphism (rs72689236) and Kawasaki disease in Taiwanese children.
Kuo HC, et al.
Journal of Human Genetics, 56(2), 161?165-161?165 (2011)
Identification of SMURF1 as a possible target for 7q21. 3?22.1 amplification detected in a pancreatic cancer cell line by in?house array?based comparative genomic hybridization.
Suzuki A, et al.
Cancer Science, 99(5), 986-994 (2008)
Guojun Yang et al.
Nanotheranostics, 7(4), 380-392 (2023-07-10)
Rationale: UV light deeply penetrates the dermis, leading to inflammation and cell death with prolonged exposure. This is a major contributor to skin photoaging. In the pharmaceutical field, fibroblast growth factors (FGFs) have gained popularity for enhancing skin quality as

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service