Skip to Content
Merck
All Photos(2)

Key Documents

SAB4503087

Sigma-Aldrich

Anti-WASP antibody produced in rabbit

affinity isolated antibody

Synonym(s):

IMD2, WAS, Wiskott-Aldrich syndrome protein, Wiskott-Aldrich syndrome protein homolog

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
conjugate:
unconjugated
application:
ELISA
IHC
WB
clone:
polyclonal
species reactivity:
human, mouse
citations:
5
technique(s):
ELISA: 1:5000
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 52 kDa

species reactivity

human, mouse

concentration

~1 mg/mL

technique(s)

ELISA: 1:5000
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... WAS(7454)

General description

Anti-WASP Antibody detects endogenous levels of total WASP protein.
WASP (Wiskott-Aldrich syndrome protein) codes for a 65 kDa protein, localized to the cytoplasm. The WASP gene is mapped to human chromosome Xp11.23.

Immunogen

The antiserum was produced against synthesized peptide derived from human WASP.

Immunogen Range: 256-305

Biochem/physiol Actions

WASP (Wiskott-Aldrich syndrome protein) is regarded as a scaffold protein that integrates cell surface signals to actin cytoskeleton. WASP gene regulates actin polymerization, contributing to T-cell activation and migration. Neural WASP is known to regulate a variety of cellular processes including pathogen infection, vesicle trafficking, invadopodium formation and growth, and neurite extension. Mutation in this gene causes WAS, a X-linked recessive immunodeficiency disorder.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Neural Wiskott-Aldrich syndrome protein (nWASP) is implicated in human lung cancer invasion
Frugtniet BA, et al.
BMC Cancer, 17(1), 224-224 (2017)
X-Linked Syndrome of Polyendocrinopathy, Immune Dysfunction, and Diarrhea Maps to Xp11.23-Xq13.3
Bennett CL, et al.
American Journal of Human Genetics, 66(2), 461-468 (2000)
Molecular difference between WASP and N-WASP critical for chemotaxis of T-cells towards SDF-1α
Jain N, et al.
Scientific Reports, 5, 15031-15031 (2015)
Novel WASP mutation in a patient with Wiskott-Aldrich syndrome: Case report and review of the literature.
Eghbali M, et al.
Allergologia et Immunopathologia, 44(5), 450-454 (2016)
Sudha Kumari et al.
The EMBO journal, 39(5), e102783-e102783 (2020-01-03)
When migratory T cells encounter antigen-presenting cells (APCs), they arrest and form radially symmetric, stable intercellular junctions termed immunological synapses which facilitate exchange of crucial biochemical information and are critical for T-cell immunity. While the cellular processes underlying synapse formation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service