Skip to Content
Merck
All Photos(2)

Key Documents

SAB4502560

Sigma-Aldrich

Anti-RAC1 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Cell migration-inducing gene 5 protein, RAC1, Ras-like protein TC25, Ras-related C3 botulinum toxin substrate 1, p21-Rac1

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 21 kDa

species reactivity

rat, human, mouse

concentration

~1 mg/mL

technique(s)

ELISA: 1:20000
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RAC1(5879)

Related Categories

General description

The gene ras-related C3 botulinum toxin substrate 1 (RAC1), with 7 exons spanning 29 kb, is mapped to human chromosome 7p22. The encoded protein belongs to the Rho family of small GTPases. RAC1 is a maternal factor expressed in decidual cells.

Immunogen

The antiserum was produced against synthesized peptide derived from human Rac1/CDC42.

Immunogen Range: 38-87

Biochem/physiol Actions

Ras-related C3 botulinum toxin substrate 1 (RAC1), expressed in decidua, modulates uterine secretions that helps in maternal-fetal communication crucial for placental development and establishment of pregnancy. RAC1 regulates the secretory activity of human endometrial stromal cells during decidualization. Members of rho family of small GTPases, including RAC1, is associated with signal transduction pathways that regulate proliferation, adhesion, and migration of cells during embryonic development and invasiveness of tumor cells. RAC1 acts as a regulator of nuclear factor-κB (NF-κB) signaling pathway, and it is also associated with inflammation.Hindrance of RAC1 expression in lens epithelial cells can be a potential method for the treatment of cataract. Elevated expression of RAC1 promotes the proliferation, migration, and invasion of lens epithelial cells, and regulates the epithelial-mesenchymal transition (EMT) process, thus, RAC1 is associated with cataract. RAC1 functions as an oncogene in tumor cells. Deletion of RAC1 retards cell migration and wound healing of epidermal keratinocytes.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rac1 Regulates Endometrial Secretory Function to Control Placental Development.
Davila J, et al.
PLoS Genetics, 11(8), e1005458-e1005458 (2015)
Small GTPase Rac1: structure, localization, and expression of the human gene.
Matos P, et al.
Biochemical and Biophysical Research Communications, 277(3), 741-751 (2000)
RAC1 overexpression promotes the proliferation, migration and epithelial-mesenchymal transition of lens epithelial cells.
Su J and Li H
International Journal of Clinical and Experimental Pathology, 8(9), 10760-10767 (2015)
Seung Hee Lee et al.
International journal of molecular sciences, 21(16) (2020-08-23)
Increased platelet activation and apoptosis are characteristic of diabetic (DM) platelets, where a Parkin-dependent mitophagy serves a major endogenous protective role. We now demonstrate that Parkin is highly expressed in both healthy platelets and diabetic platelets, compared to other mitochondria-enriched

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service